Skip to Content
Merck
All Photos(3)

Key Documents

MABE1013

Sigma-Aldrich

Anti-RRP8 Antibody, clone 5C7.1

clone 5C7.1, from mouse

Synonym(s):

Ribosomal RNA-processing protein 8, Cerebral protein 1, Nucleomethylin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5C7.1, monoclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

General description

Ribosomal RNA-processing protein 8 (UniProt: O43159; also known as Cerebral protein 1, Nucleomethylin) is encoded by the RRP8 (also known as KIAA0409, NML, hucep-1) gene (Gene ID: 23378) in human. Ribosome biosynthesis is one of the most energy-consuming cellular process, which adapts to changes in intracellular energy status. Ribosomal RNA-processing protein 8 is a novel nuclear protein that serves as an essential component of the energy-dependent nucleolar silencing) complex (eNoSC) that mediates silencing of rDNA in response to intracellular energy status and acts by recruiting histone-modifying enzymes. The eNoSC complex is able to sense the energy status of cell: upon glucose starvation or upon elevation of NAD+/NADP+ ratio and activates SIRT1 that leads to deacetylation of histone H3 followed by dimethylation of H3 at Lysine 9 (H3K9me2) by SUV39H1 and the formation of silent chromatin in the rDNA locus. In the complex, RRP8 binds to H3K9me2 and probably acts as a methyltransferase.

Specificity

Clone 5C7.1 specifically detects Ribosomal RNA-processing protein 8. It targets an epitope with in the C-terminal half.

Immunogen

GST-tagged recombinant fragment corresponding to 98 amino acids from the C-terminal half of human Ribosomal RNA-processing protein 8.

Application

Anti-RRP8, clone 5C7.1, Cat. No. MABE1013, is a highly specific mouse monoclonal antibody, that targets Ribosomal RNA-processing protein 8 and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected RRP8 in HeLa, A431, HUVEC, and NIH/3T3 cells.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected RRP8 in HeLa cell lysate.

Target description

~53 kDa observed; 50.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service