Skip to Content
Merck
All Photos(1)

Key Documents

MABC1152

Sigma-Aldrich

Anti-LINE-1 ORF1p Antibody, clone 4H1

clone 4H1, from mouse

Synonym(s):

LINE-1 retrotransposable element ORF1 protein, L1ORF1p, LINE retrotransposable element 1, LINE1 retrotransposable element 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4H1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... L1RE1 (4029)

General description

LINE-1 retrotransposable element ORF1 protein (UniProt: Q9UN81; also known L1ORF1p, LINE retrotransposable element 1, LINE1 retrotransposable element 1) is encoded by the LIRE1 (also known as LRE1) gene in human. LINE1-ORF 1P belongs to the transposase 22 family. It is mainly cytoplasmic protein and is enriched in stress granules, but may rarely be detected in the nucleolus. It contains a coiled coil domain that mediates its homotrimerization. The RNA recognition motif (RRM) the C-terminal domain are required for its proper RNA-binding activity. LINE1-ORF 1P is a nucleic acid-binding protein which is essential for retrotransposition of LINE-1 elements in the genome. It is also reported to function as a nucleic acid chaperone binding its own transcript and therefore preferentially mobilizing the transcript from which they are encoded. Retrotransposition is a shown to be a complex process that involves transcription of the LINE1, transport of its RNA to the cytoplasm, translation of the bicistronic RNA, formation of a ribonucleoprotein particle, its re-import to the nucleus and then target-primed reverse transcription at the integration site. Somatic LINE1 insertions are rare in most tissues, but are found in brain, testis, and many cancer types and have been implicated in carcinogenesis. Its expression is consistently detected in p53 mutant tumors, but, little or no expression is detected in Wilms tumors that were normal for p53. (Ref.: Goodier, JL et al. (2013). Nucleic Acids Res. 41(15): 7401-7419).

Specificity

Clone 4H1 specifically detects LINE-1 ORF1 protein in MCF7 cells. This clone targets the sequence corresponding to 10 amino acids from the N-terminal region of LINE-1 ORF1p.

Immunogen

A linear peptide corresponding to 10 amino aicds from the N-terminal region of human LINE-1 ORF1 protein.

Application

Detect LINE-1 retrotransposable element ORF1 protein using this mouse monoclonal Anti-LINE-1 ORF1p Anibody, clone 4H1, Cat. No. MABC1152. It is used in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected LINE-1 ORF1p in Immunohistochemistry applications (Rodic, N., et. al. (2014). AM J Pathol. 184(5):1280-6; Doucet-O′Hare, T.T., et. al. (2015). Proc Natl Acad Sci USA. 112(35):E4894-900; Wylie, A., et. al. (2016). Genes Dev. 30(1):64-77).

Immunocytochemistry Analysis: A representative lot detected LINE-1 ORF1p in Immunocytochemistry applications (Taylor, M.S., et. al. (2013). Cell 155(5):1034-48).

Immunoprecipitation Analysis: A representative lot detected LINE-1 ORF1p in Immunoprecipitation applications (Taylor, M.S., et. al. (2013). Cell 155(5):1034-48).

Western Blotting Analysis: A representative lot detected LINE-1 ORF1p in Western Blotting applications (Taylor, M.S., et. al. (2013). Cell 155(5):1034-48).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LINE-1 ORF1p in 10 µg of MCF7 cell lysate.

Target description

~42 kDa observed; 40.06 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gavin C Pereira et al.
Mobile DNA, 9, 35-35 (2018-12-20)
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disease involving loss of motor neurons and having no known cure and uncertain etiology. Several studies have drawn connections between altered retrotransposon expression and ALS. Certain features of the LINE-1 (L1) retrotransposon-encoded
Adam Idica et al.
The Journal of biological chemistry, 292(50), 20494-20508 (2017-10-05)
Repetitive elements, including LINE-1 (L1), comprise approximately half of the human genome. These elements can potentially destabilize the genome by initiating their own replication and reintegration into new sites (retrotransposition). In somatic cells, transcription of L1 elements is repressed by
Paul A Roberson et al.
Journal of applied physiology (Bethesda, Md. : 1985), 127(4), 895-904 (2019-08-02)
The long interspersed nuclear element-1 (L1) is a retrotransposon that constitutes 17% of the human genome and is associated with various diseases and aging. Estimates suggest that ~100 L1 copies are capable of copying and pasting into other regions of
Daniel Ardeljan et al.
Clinical chemistry, 63(4), 816-822 (2017-02-12)
A large portion of intronic and intergenic space in our genome consists of repeated sequences. One of the most prevalent is the long interspersed element-1 (LINE-1, L1) mobile DNA. LINE-1 is rightly receiving increasing interest as a cancer biomarker. Intact
Nian Liu et al.
Nature, 553(7687), 228-232 (2017-12-07)
Transposable elements, also known as transposons, are now recognized not only as parasitic DNA, the spread of which in the genome must be controlled by the host, but also as major players in genome evolution and regulation. Long interspersed element-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service