Skip to Content
Merck
All Photos(2)

Key Documents

MAB5466

Sigma-Aldrich

Anti-Bestrophin Antibody

Chemicon®, from mouse

Synonym(s):

Anti-Anti-ARB, Anti-Anti-BEST, Anti-Anti-BMD, Anti-Anti-Best1V1Delta2, Anti-Anti-RP50, Anti-Anti-TU15B, Anti-Anti-VMD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

monkey, pig, mouse, bovine, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BEST1(7439)

Specificity

Reacts with bestrophin.

Immunogen

Recombinant human bestrophin protein.

Application

Detect Bestrophin using this Anti-Bestrophin Antibody validated for use in WB, IH.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western blot: 5 μg/mL of this antibody detected Bestrophin in bovine retina cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:20 dilution from a representative lot detected Bestrophin in mouse retina tissue sections.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.08% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Petrus-Reurer et al.
Stem cells translational medicine, 9(8), 936-953 (2020-04-23)
As pluripotent stem cell (PSC)-based reparative cell therapies are reaching the bedside, there is a growing need for the standardization of studies concerning safety of the derived products. Clinical trials using these promising strategies are in development, and treatment for
Biju B Thomas et al.
Frontiers in neuroscience, 15, 752958-752958 (2021-11-13)
End-stage age-related macular degeneration (AMD) and retinitis pigmentosa (RP) are two major retinal degenerative (RD) conditions that result in irreversible vision loss. Permanent eye damage can also occur in battlefields or due to accidents. This suggests there is an unmet
Altered zinc homeostasis in a primary cell culture model of the retinal pigment epithelium.
??lvarez-Barrios, et al.
Frontiers in nutrition, 10, 1124987-1124987 (2023)
Teisha J Rowland et al.
Journal of tissue engineering and regenerative medicine, 7(8), 642-653 (2012-04-20)
A potential application of human embryonic stem cells (hESCs) and induced pluripotent stem cells (iPSCs) is the generation of retinal pigmented epithelium (RPE) to treat age-related macular degeneration (AMD), a common but incurable retinal disease. RPE cells derived from hESCs
Divya Sinha et al.
American journal of human genetics, 107(2), 278-292 (2020-07-25)
Dominantly inherited disorders are not typically considered to be therapeutic candidates for gene augmentation. Here, we utilized induced pluripotent stem cell-derived retinal pigment epithelium (iPSC-RPE) to test the potential of gene augmentation to treat Best disease, a dominant macular dystrophy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service