Skip to Content
Merck
All Photos(1)

Key Documents

MAB5350

Sigma-Aldrich

Anti-Choline Acetyltransferase Antibody

CHEMICON®, mouse monoclonal, 28C4

Synonym(s):

ChAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Choline Acetyltransferase Antibody, clone 28C4, clone 28C4, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

28C4, monoclonal

species reactivity

human, rat, guinea pig, monkey

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHAT(1103)
rat ... Chat(290567)
rhesus monkey ... Chat(709977)

Specificity

Reacts with Choline Acetyltransferase (ChAT).

Immunogen

Purified human placenta ChAT.

Application

Anti-Choline Acetyltransferase Antibody, clone 28C4 detects level of Choline Acetyltransferase & has been published & validated for use in ELISA, IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers
Western blotting: 1:1,000-1:10,000 using a high sensitivity ECL kit. Recognizes a protein of ~67 kDa. May also recognize a protein of 35 kDa which is thought to be a degradation product. Suggested dilution buffer is TBS containing 5% skim milk powder and 0.05% Tween 20.

Immunohistochemistry on free-floating frozen sections of Rhesus monkey brain fixed with 4% paraformaldehyde and coronal vibratome sections of rat brain fixed with 4% paraformaldehyde and 0.2% glutaraldehyde.

ELISA (two side)

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaoqin Huang et al.
Molecular medicine reports, 11(5), 3285-3290 (2015-01-17)
The present study was conducted to investigate the 11C‑Choline metabolic mechanism and examine the association between 11C‑Choline metabolism and uptake in different pathological types of lung cancer. A total of 18 tumor specimens and corresponding normal lung tissues were collected
Katarina Zvarova et al.
The Journal of comparative neurology, 489(4), 501-517 (2005-07-19)
We examined the distribution and fate of cocaine- and amphetamine-regulated transcript peptide (CARTp)(55-102)-immunoreactive (IR) structures in the neonatal and adult rat urinary bladder. Double-labeling studies examining CARTp with tyrosine hydroxylase (TH), neuronal nitric oxide synthase (nNOS), or choline acetyltransferase (ChAT)
ARG1-expressing microglia show a distinct molecular signature and modulate postnatal development and function of the mouse brain.
Stratoulias, et al.
Nature Neuroscience, 26, 1008-1020 (2023)
Hanqing Tang et al.
International heart journal, 62(3), 647-657 (2021-05-18)
Bioinformatics analysis showed that miR-448-5p expression in the myocardial tissue of rats with myocardial infarction significantly increased, suggesting that it may participate in myocardial cell apoptosis in myocardial infarction. This study aimed to explore the protective effects of miR-448-5p on
C M Hernandez et al.
The Journal of pharmacology and experimental therapeutics, 316(2), 679-694 (2005-10-11)
This study was designed to determine 1) whether repeated exposures to the acetylcholinesterase inhibitors (AChEIs) galantamine (GAL) or donepezil (DON) resulted in positive effects on nerve growth factor (NGF) and its receptors, cholinergic proteins, and cognitive function in the aged

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service