Skip to Content
Merck
All Photos(2)

Documents

MAB305

Sigma-Aldrich

Anti-Choline Acetyltransferase Antibody, clone 1E6

ascites fluid, clone 1E6, Chemicon®

Synonym(s):

ChAT, Choline Acetylase, CHOACTase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1E6, monoclonal

species reactivity

human, rat, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CHAT(1103)
rat ... Chat(290567)
rhesus monkey ... Chat(709977)

Specificity

Recognizes cholinergic neurons in the brain and spinal cord (CNS).

Immunogen

Choline acetyltransferase purified from rat brain.

Application

Detect Choline Acetyltransferase using this Anti-Choline Acetyltransferase Antibody, clone 1E6 validated for use in IH.
Immunohistochemistry: 1:100-1:250. See immunohistochmistry procedure below.

Optimal working dilutions must be determined by the end user.

IMMUNOHISTOCHEMISTRY PROCEDURE (PAP TECHNIQUE) FOR MAB305, MONOCLONAL ANTIBODY TO CHOLINE ACETYLTRANSFERASE

I) Perfusion & Sectioning Procedure

1. Perfuse through the heart with a fixative solution containing 4% paraformaldehyde in 0.12 M phosphate buffer (pH 7.3) for light microscopy (LM), and additionally, 0.1% gluteraldehyde and .002% CaCl2 for electron microscopy (EM).

2. Remove brain and postfix 2-18 hours at 4°C in 4% paraformaldehyde in 0.12 M phosphate buffer.

3. After brain is blocked for sectioning, wash in several changes of buffer for 2-3 hours.

4. Specimens for EM are sectioned on a Vibratome (50 μm) and rinsed in buffer, those for LM should be cryoprotected in 30% sucrose in buffer.

5. After freezing with dry ice, 30-40 μm thick sections of LM specimens are cut on a cryostat.

6. Sections are rinsed, and then stored in phosphate buffer containing 0.1% sodium azide.

II) Staining Procedure

Tissue is processed as freely-floating sections in continuously agitated solutions. All incubations are performed at room temperature unless otherwise stated.

1.a. For localizing ChAT-positive somata and dendrites:

Sections are washed in 0.1 M Tris-buffered saline (TBS; containing 1.4% NaCl, pH 7.3) only. No detergent or enzyme pretreatment is used.

b. For localizing ChAT-positive terminal-like structures:

Incubate sections in TBS (pH 8.1) for 5 minutes at 37°C. Transfer sections to TBS (pH 8.1) containing pronase (1.2 μg/mL) for 1 1/2-2 minutes at 37°C, followed by several ice cold buffer washes for a total of 5 minutes. The concentration of pronase and incubation time of the digestion should be evaluated for each region examined.

c. For localizing ChAT immunoreactivity and subsequently counterstaining the sections:

Incubation in TBS containing 0.1%-0.8% Triton X-100 for 15 minutes may increase the tissue penetration of the immunoreagents, but it also raises the background staining.

2. Incubate sections in normal goat serum (3-5%) for one hour. The working solutions of all antisera should also contain similarly diluted normal goat serum.

3. Incubate in anti-ChAT monoclonal antibody solution (Suggested working dilution 1:250, final working dilution must be determined by end user) for 2 hours at room temperature and then for an additional 6-18 hours at 4°C.

4. Incubate with second antibody (i.e. Goat anti-Mouse IgG, Cat. No.: AP124, dilution 1:50-100) for 1-2 hours.

5. Incubate with diluted PAP complex (i.e. Mouse PAP, Cat No.: PAP14, conc. 25-50 μg/mL) for one hour.

6. After rinsing in buffer, the second antibody and PAP steps are repeated for 40 minutes to 1 hour each in order to amplify staining intensity, particularly of small ChAT-containing structures.

7. React for 15 minutes with 0.06% 3,3′-diaminobenzidine×4 HCl (DAB; diluted in phosphate buffered saline, pH 7.3) and 0.006% H2O2.

8. Specimens for routine LM are postfixed for 1 minutes in 0.005% OsO4 (osmium tetraoxide), and then mounted, dehydrated and coverslipped. Selected regions blocked for EM are postfixed in 2% OsO4 for 1 hour, en bloc stained with uranyl acetate, and flat-embedded in Epon-Araldite resin.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Physical form

Ascites fluid containing no preservatives.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Alex Goddard et al.
Journal of neurophysiology, 98(6), 3486-3493 (2007-09-28)
Cholinergic neurons in the parabigeminal nucleus of the rat midbrain were studied in an acute slice preparation. Spontaneous, regular action potentials were observed both with cell-attached patch recordings as well as with whole cell current-clamp recordings. The spontaneous activity of
Tamunotonye Omoluabi et al.
Alzheimer's & dementia (New York, N. Y.), 7(1), e12231-e12231 (2022-01-11)
The earliest abnormality associated with Alzheimer's disease (AD) is the presence of persistently phosphorylated pretangle tau in locus coeruleus (LC) neurons. LC neuron numbers and fiber density are positive predictors of cognition prior to death. Using an animal model of
Chondroitinase applied to peripheral nerve repair averts retrograde axonal regeneration.
Graham, JB; Neubauer, D; Xue, QS; Muir, D
Experimental neurology null
Characterization of cognitive deficits in rats overexpressing human alpha-synuclein in the ventral tegmental area and medial septum using recombinant adeno-associated viral vectors.
Hall, H; Jewett, M; Landeck, N; Nilsson, N; Schagerlof, U; Leanza, G; Kirik, D
Testing null
Samantha R Eck et al.
Biological psychiatry, 88(7), 566-575 (2020-07-01)
Stress exacerbates symptoms of schizophrenia and attention-deficit/hyperactivity disorder, which are characterized by impairments in sustained attention. Yet how stress regulates attention remains largely unexplored. We investigated whether a 6-day variable stressor altered sustained attention and the cholinergic attention system in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service