Skip to Content
Merck
All Photos(1)

Key Documents

MAB3868

Sigma-Aldrich

Anti-DNA Antibody, single stranded

clone TNT-3, Chemicon®, from mouse

Synonym(s):

ssDNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TNT-3, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1, kappa

shipped in

dry ice

target post-translational modification

unmodified

Related Categories

Specificity

Reacts with single stranded DNA (ssDNA). The antibody is useful for staining nucleated cells. The antibody can also be used to identify necrotic regions of tumors and damaged tissues. By immunohistochemistry, stains heterochromatic regions of cell nucleus.

Immunogen

Human Raji cell nuclei

Application

Detect DNA using this Anti-DNA Antibody, single stranded validated for use in ELISA, FC, IC, IH(P).
Immunocytochemistry on paraformaldehyde fixed cells

Immunohistochemistry on paraformaldehyde or B5 fixed tissue sections.

Flow cytometry: 5-10 mg/mL using 2% paraformaldehyde fixed cells.

ELISA

Optimal working dilutions must be determined by end user.

STAINING PATTERN:

Stains heterochromatic regions of cell nucleus.
Research Category
Epitope Tags & General Use
Research Sub Category
Organelle & Cell Markers

Physical form

Format: Purified
Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months from date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Nannini et al.
mAbs, 13(1), 1864084-1864084 (2021-01-01)
Phage display technology in combination with next-generation sequencing (NGS) currently is a state-of-the-art method for the enrichment and isolation of monoclonal antibodies from diverse libraries. However, the current NGS methods employed for sequencing phage display libraries are limited by the
LiJun Wang et al.
Cellular reprogramming, 23(2), 99-107 (2021-04-17)
Zygotic epigenetic reprogramming is the major initial event in embryo development to acquire a totipotent potential. However, the patterns of epigenetic modifications in bovine zygote were not well clarified, especially in the first cell cycle of bovine somatic cell nuclear
Fan Guo et al.
Cell stem cell, 15(4), 447-459 (2014-09-16)
The epigenomes of mammalian sperm and oocytes, characterized by gamete-specific 5-methylcytosine (5mC) patterns, are reprogrammed during early embryogenesis to establish full developmental potential. Previous studies have suggested that the paternal genome is actively demethylated in the zygote while the maternal
New histone supply regulates replication fork speed and PCNA unloading.
Mejlvang, J; Feng, Y; Alabert, C; Neelsen, KJ; Jasencakova, Z; Zhao, X; Lees, M; Sandelin et al.
The Journal of cell biology null
Paula Andrea Marin et al.
Frontiers in cell and developmental biology, 8, 602956-602956 (2021-01-09)
DNA double-strand breaks (DSBs) are among the most deleterious lesions that threaten genome integrity. To address DSBs, eukaryotic cells of model organisms have evolved a complex network of cellular pathways that are able to detect DNA damage, activate a checkpoint

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service