Skip to Content
Merck
All Photos(1)

Key Documents

MAB3864

Sigma-Aldrich

Anti-DNA/Histone H1 Antibody

Chemicon®, from mouse

Synonym(s):

Anti-H1.4, Anti-H1E, Anti-H1F4, Anti-H1s-4, Anti-HIST1H1E, Anti-RMNS, Anti-dJ221C16.5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H1-1(3024)

Specificity

Reacts with DNA/Histone 1. The antibody can be used to stain nuclei of all species and to identify necrosis. By immunohistochemistry, the antibody shows dense staining of nuclei with no staining of nucleoli observed.

Immunogen

Human Raji cell nuclei

Application

Anti-DNA/Histone H1 Antibody is a high quality Mouse Monoclonal Antibody for the detection of DNA/Histone H1 & has been validated in ELISA, FC, ICC & IHC.
Immunocytochemistry: 5-10 μg/mL on 2% paraformaldehyde fixed cells (10 minutes at room temperature)

Immunohistochemistry: 5-10 μg/mL on paraformaldehyde or B5 fixed tissue sections.

Flow cytometry: 5-10 μg/mL using 2% paraformaldehyde fixed cells.

ELISA, direct and indirect

Optimal working dilutions must be determined by end user.

STAINING PATTERN:

Nuclear staining observed. Dense staining of nuclei. No staining of nucleoli.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Physical form

Format: Purified
Protein A purified culture supernatant. Liquid in PBS

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months from date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Liu et al.
Mucosal immunology, 12(4), 862-873 (2019-04-13)
Extra-intestinal manifestations (EIM) are common in inflammatory bowel disease (IBD). One such EIM is sub-clinical pulmonary inflammation, which occurs in up to 50% of IBD patients. In animal models of colitis, pulmonary inflammation is driven by neutrophilic infiltrations, primarily in
Simona Grasso et al.
Thrombosis research, 161, 36-42 (2017-11-28)
The circulating zymogen form of Factor VII activating protease (FSAP) can be activated by histones and nucleosomes in vivo. These cell-death-associated nuclear factors are also actively extruded into the extracellular space by neutrophils through a process called neutrophil extracellular trap
Verena Peek et al.
Pharmaceuticals (Basel, Switzerland), 14(6) (2021-07-03)
High mobility group box (HMGB)1 action contributes to late phases of sepsis, but the effects of increased endogenous plasma HMGB1 levels on brain cells during inflammation are unclear. Here, we aimed to further investigate the role of HMGB1 in the
Nicole de Buhr et al.
Frontiers in microbiology, 9, 966-966 (2018-06-06)
Granulocytes mediate the first line of defense against infectious diseases in humans as well as animals and they are well known as multitasking cells. They can mediate antimicrobial activity by different strategies depending on the pathogen they encounter. Besides phagocytosis
Nicole de Buhr et al.
Cellular microbiology, 19(2) (2016-07-28)
Streptococcus suis is an important meningitis-causing pathogen in pigs and humans. Neutrophil extracellular traps (NETs) have been identified as host defense mechanism against different pathogens. Here, NETs were detected in the cerebrospinal fluid (CSF) of S. suis-infected piglets despite the presence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service