Skip to Content
Merck
All Photos(6)

Key Documents

MAB360

Sigma-Aldrich

Anti-Glial Fibrillary Acidic Protein Antibody

CHEMICON®, mouse monoclonal, GA5

Synonym(s):

GFAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

GA5, monoclonal

species reactivity

rat, human, mouse, rabbit, chicken, pig, bovine

packaging

antibody small pack of 25 μL

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GFAP(2670)

General description

Glial fibrillary acidic protein is a class-III intermediate filament. GFAP is the main constituent of intermediate filaments in astrocytes and serves as a cell specific marker that distinguishes differentiated astrocytes from other glial cells during the development of the central nervous system.

Specificity

Glial fibrillar acidic protein. On western blots of extracts from a human glioma cell line (U33CG/343MG), MAB360 recognizes a band at approximately 51 kDa corresponding to GFAP (Debus, 1983). By immunohistochemistry it recognizes astrocytes and Bergmann glia cells, glioma and glial cell derived tumors. Shows no cross-reactivity with vimentin.
Reactivity with other species has not been determined.

Immunogen

Purified GFAP from porcine spinal cord.

Application

Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5 detects level of Glial Fibrillary Acidic Protein & has been published & validated for use in IC, IH, IH(P) & WB with more than 65 product citations.
Immunocytochemistry:
A previous lot was used on cultured mammalian cells.

Immunohistochemistry(paraffin):
1:400-1:800 dilution from a previous lot was used on alcohol-fixed paraffin embedded sections of rat brain (cerebrum or cerebellum) and human brain.

Immunoblotting: A 1:1000 dilution of a previous lot was used.

Optimal working dilutions must be determined by the end user

Quality

Routinely evaluated by Western Blot on Mouse brain lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected GFAP on 10 μg of Mouse brain lysates.

Target description

~ 51 kDa

Linkage

Replaces: 04-1031; 04-1062

Physical form

Unpurified mouse monoclonal IgG1 liquid in buffer containing 15 mM sodium azide.

Analysis Note

Control
Positive Control: Cultured astrocytes, brain tissue (white matter), spinal chord, retina

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lateral fluid percussion: model of traumatic brain injury in mice.
Alder, J; Fujioka, W; Lifshitz, J; Crockett, DP; Thakker-Varia, S
Journal of Visualized Experiments null
Y Jiang et al.
Brain research, 1332, 100-109 (2010-03-30)
Recent work suggests that diabetes affects processing of peripheral, spinal and supraspinal signals in the spinal cord. However, there is little evidence for spinal cord lesions that would account for alterations in behavioral responses induced by experimental diabetes. Therefore, we
Adenosine kinase as a target for therapeutic antisense strategies in epilepsy.
Theofilas, P; Brar, S; Stewart, KA; Shen, HY; Sandau, US; Poulsen, D; Boison, D
Epilepsia null
Biochip?laser cell deposition system to assess polarized axonal growth from single neurons and neuron?glia pairs in microchannels with novel asymmetrical geometries.
Pirlo, RK; Sweeney, AJ; Ringeisen, BR; Kindy, M; Gao, BZ
Biomicrofluidics null
J C Durham-Lee et al.
Neuroscience, 202, 454-464 (2011-10-25)
Angiopoietin-1 (Ang-1) and angiopoietin-2 (Ang-2) have opposing effects on blood vessels, with Ang-2 being mainly induced during the endothelial barrier breakdown. It is known that spinal cord injury (SCI) induces lasting decreases in Ang-1 levels, underlying endothelial barrier disruption, but

Protocols

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Questions

  1. Can this antibody be used in IHC frozen(IHC-F) rather than IHC paraffin(IHC-P)?

    1 answer
    1. This material has not been tested for suitability in the application for IHC-F.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service