Skip to Content
Merck
All Photos(8)

Key Documents

AB5804

Sigma-Aldrich

Anti-Glial Fibrillary Acidic Protein Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

glial fibrillary acidic protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, bovine, canine, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GFAP(2670)

Related Categories

General description

Glial fibrillary acidic protein is a class-III intermediate filament. GFAP is the main constituent of intermediate filaments in astrocytes and serves as a cell specific marker that distinguishes differentiated astrocytes from other glial cells during the development of the central nervous system.

Specificity

It is expected that the antibody will react with most mammalian GFAP due to homology. Reactivity with other species has not been confirmed.
Recognizes Glial Fibrillary Acidic Protein (GFAP).

Immunogen

Purified bovine GFAP

Application

Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody is an antibody against Glial Fibrillary Acidic Protein (GFAP) for use in IC, IH, IH(P) with more than 65 product citations.
Immunohistochemistry(paraffin):
Representative images from a previous lot.

Immunocytochemistry:
A 1:1,000 dilution of a previous lot was used in Immunocytochemistry.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Quality

Routinely evaluated by immunohistochemistry on brain tissue, astrocytes, and neurons.

Immunohistochemistry(paraffin) Analysis:
GFAP (IHC2078) staining of Human Brain. Tissue pretreated with Citrate Buffer, pH 6.0. Pre-diluted polyclonal antibody, using IHC-Select Detection with HRP-DAB. Glial cells stain strongly (brown).
Optimal Staining with Citrate Buffer Epitope Retrieval: Human Brain

Target description

50 kDa

Linkage

Replaces: AB1540

Physical form

Rabbit serum. Liquid. Contains no preservative.
Unpurified

Storage and Stability

Stable for 1 year at -20°C undiluted aliquots from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Mouse brain tissue, rat astrocytes, rat neurons.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diego Mastroeni et al.
PloS one, 4(8), e6617-e6617 (2009-08-13)
DNA methylation is capable of modulating coordinate expression of large numbers of genes across many different pathways, and may therefore warrant investigation for their potential role between genes and disease phenotype. In a rare set of monozygotic twins discordant for
Congenital anomalies and rhabdoid tumor associated with 22q11 germline deletion and somatic inactivation of the SMARCB1 tumor suppressor.
Toth, G; Zraly, CB; Thomson, TL; Jones, C; Lapetino, S; Muraskas, J; Zhang, J; Dingwall, AK
Genes Chromosomes Cancer null
A splice variant of the Wilms' tumour suppressor Wt1 is required for normal development of the olfactory system.
Wagner, N; Wagner, KD; Hammes, A; Kirschner, KM; Vidal, VP; Schedl, A; Scholz, H
Development null
Antonio Currais et al.
Aging cell, 11(6), 1017-1026 (2012-09-04)
Mounting evidence supports a link between diabetes, cognitive dysfunction, and aging. However, the physiological mechanisms by which diabetes impacts brain function and cognition are not fully understood. To determine how diabetes contributes to cognitive dysfunction and age-associated pathology, we used
Jack T Mosher et al.
Developmental biology, 303(1), 1-15 (2006-11-23)
We have systematically examined the developmental potential of neural crest stem cells from the enteric nervous system (gut NCSCs) in vivo to evaluate their potential use in cellular therapy for Hirschsprung disease and to assess differences in the properties of

Articles

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

ReNcell neural progenitors are immortalized human neural stem cell lines that can differentiate into neurons, astrocytes sand oligodendrocytes.

Protocols

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service