Skip to Content
Merck
All Photos(1)

Key Documents

MAB3199

Sigma-Aldrich

Anti-E-Cadherin Antibody, clone 67A4

clone 67A4, Chemicon®, from mouse

Synonym(s):

CD324

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

67A4, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CDH1(999)

Specificity

Antibodies against E-cadherin are suitable to characterize epithelial cells. E-Cadherin is involved in adhesion of epithelial cells (mostly homotypic and homophilic). E-cadherin is also expressed on immature erythroid cells. The antibody 67A4 is suitable to study E-cadherin expression by flow cytometry. In addition, it blocks the interaction of E-cadherin molecules and therefore is suitable to study E-cadherin-mediated signal transduction.

Antigen is expressed in non-neural epithelial tissues.

Immunogen

T-47D breast carcinoma line

Application

Anti-E-Cadherin Antibody, clone 67A4 detects level of E-Cadherin & has been published & validated for use in FC & WB.
Immunoblotting: 123kD in membrane fractions; MCF-7 membranes positive control. Confluent cells were washed three times with ice-cold PBS and lysed in 50mM HEPES pH 7.5, 150mM NaCl, 1mM EDTA, 10% glycerine, 1% triton X-100, 20mM Na(4)P(2)O(7), 2mM Na-vanadate, 100mM NaF, 2mM PMSF, 20μg/mL leupeptin, 1mM pNPP(para-nitrophenylphosphate), 20μg/mL aprotinin, and 200μM NH(4)-molybdate. Crude extracts were clarified by centrifugation (10,000 x g 10 min), and supernatants were taken for immunoprecipitation. Protein concentration of cleared lysates were determined by BCA protein assay. {Buhring, et al Luekemia 10:106-116, 1996}.



FACS Analysis

Functional blocking of E-cadherin

Does not work well for immunoprecipitation

Original paper, Buhring, H et al Leukemia 10:106-116, 1996.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Linkage

Replaces: 04-1103

Physical form

Format: Purified
Protein A purified immunoglobulin. Liquid in 0.02M PBS with NaCl, pH=7.6, 0.1% Sodium azide.

Storage and Stability

Maintain 2-8°C in undiluted aliquots for up to 12 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Evans et al.
Gene therapy, 22(10), 802-810 (2015-05-26)
Prostate cancer is the most common cancer in men of the western world. To date, no effective treatment exists for metastatic prostate cancer and consequently, there is an urgent need to develop new and improved therapeutics. In recent years, the
Claudia M García-Cuellar et al.
International journal of molecular sciences, 22(23) (2021-12-11)
Air pollution presents a major environmental problem, inducing harmful effects on human health. Particulate matter of 10 μm or less in diameter (PM10) is considered an important risk factor in lung carcinogenesis. Epithelial-mesenchymal transition (EMT) is a regulatory program capable
Androgen receptor-independent function of FoxA1 in prostate cancer metastasis.
Jin, HJ; Zhao, JC; Ogden, I; Bergan, RC; Yu, J
Cancer Research null
Li Li et al.
Biophysical journal, 98(11), 2442-2451 (2010-06-02)
Clonality is, at present, the only means by which the self-renewal potential of a given stem cell can be determined. To assess the clonality of human embryonic stem cells (hESC), a protocol involving seeding wells at low cell densities is
Kai-Yen Peng et al.
Frontiers in oncology, 11, 716055-716055 (2021-09-28)
Over 90% of colorectal cancer (CRC) patients have mutations in the Wnt/β-catenin pathway, making the development of biomarkers difficult based on this critical oncogenic pathway. Recent studies demonstrate that CRC tumor niche-stromal cells can activate β-catenin in cancer-initiating cells (CICs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service