Skip to Content
Merck
All Photos(1)

Documents

MAB2503

Sigma-Aldrich

Anti-Elastin Antibody, clone 10B8

clone 10B8, Chemicon®, from mouse

Synonym(s):

Anti-ADCL1, Anti-SVAS, Anti-WBS, Anti-WS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10B8, monoclonal

species reactivity

mouse, bovine, human, chicken

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ELN(2006)

Specificity

Recognizes human Elastin. The antibody is also reactive with elastin from chicken, and bovine and mouse. Antibody specificity has been determined by immunoblotting of purified chick tropoelastin and immunoprecipitation of tritium-labeled chick tropoelastin from extracts of chick aorta organ cultures.

Immunogen

Tropoelastin from 17-day old lathyric chick embryo aortae, extracted an purified according to the method of Rich and Foster (1982) Meth. Enzymol. 82: 665-673.

Application

Anti-Elastin Antibody, clone 10B8 detects level of Elastin & has been published & validated for use in ELISA, IF, IP & WB.
Immunoblotting

Immunofluorescence

Immunoprecipitation

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Target description

66 kDa

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Skin

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas J Kean et al.
Connective tissue research, 57(4), 319-333 (2016-04-30)
To determine the effect of reduced (5%) oxygen tension on chondrogenesis of auricular-derived chondrocytes. Currently, many cell and tissue culture experiments are performed at 20% oxygen with 5% carbon dioxide. Few cells in the body are subjected to this supra-physiological
Long-term stability and safety of transgenic cultured epidermal stem cells in gene therapy of junctional epidermolysis bullosa.
De Rosa, L; Carulli, S; Cocchiarella, F; Quaglino, D; Enzo, E; Franchini, E; Giannetti et al.
Stem Cell Reports null
Léonard Samain-Aupic et al.
Scientific reports, 13(1), 10550-10550 (2023-06-30)
Tactile sensitivity generally decreases with aging and is associated with impairments in skin properties. Products that hydrate the skin can combat touch deficits and aromatic compounds have been shown to improve skin mechanical properties. Thus, we tested a base cosmetic
Kathleen R Chirco et al.
Acta biomaterialia, 57, 293-303 (2017-05-10)
Endothelial cells (ECs) of the choriocapillaris are one of the first cell types lost during age-related macular degeneration (AMD), and cell replacement therapy is currently a very promising option for patients with advanced AMD. We sought to develop a reliable
Wei Chen et al.
Frontiers in bioengineering and biotechnology, 11, 1135936-1135936 (2023-06-07)
The extracellular matrix (ECM) is assembled by hundreds of proteins orchestrating tissue patterning and surrounding cell fates via the mechanical-biochemical feedback loop. Aberrant ECM protein production or assembly usually creates pathological niches eliciting lesions that mainly involve fibrogenesis and carcinogenesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service