Skip to Content
Merck
All Photos(1)

Key Documents

MAB1961

Sigma-Aldrich

Anti-Integrin αVβ5 Antibody

CHEMICON®, mouse monoclonal, P1F6

Synonym(s):

Anti-CD51, Anti-MSK8, Anti-VNRA, Anti-VTNR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin αVβ5 Antibody, clone P1F6, clone P1F6, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P1F6, monoclonal

species reactivity

human, chicken

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

suitability

not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

General description

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha V chain. Alpha chain V undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.

Specificity

Reacts with the human integrin alphaVbeta5.

Immunogen

Human UCLA P3 cells (lung carcinoma)

Application

Inhibition of attachment of HT29 cells to vitronectin: >1:1,000

Inhibition of attachment of carcinoma and melanoma cells to vitronectin.

Immunohistochemistry: for use on acetone or paraformaldehyde fixed tissue. (Staining of formalin-fixed tissues REQUIRES boiling tissue sections in 1mM EDTA, pH 8.0, for 10-20 min followed by cooling at RT for 20 min.)

Immunocytochemistry: 1:1500. For use on human carcinomas, normal epithelium and keratinocytes (Acetone Fixed Only).

Immunoprecipitation

FACS

EIA

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Integrin αVβ5 Antibody, clone P1F6 is validated for use in FC, IC, IH, IP, FUNC for the detection of Integrin αVβ5.

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Epithelium, keratinocytes

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of Hsp27 affects the metastatic phenotype of human melanoma cells in vitro.
Aldrian, S; Trautinger, F; Frohlich, I; Berger, W; Micksche, M; Kindas-Mugge, I
Cell Stress & Chaperones null
The differentiation of pancreatic tumor-initiating cells by vitronectin can be blocked by cilengitide.
Cabarcas, SM; Sun, L; Mathews, L; Thomas, S; Zhang, X; Farrar, WL
Pancreas null
Ba Reun Kim et al.
Biochimica et biophysica acta, 1842(9), 1324-1332 (2014-05-20)
Periostin, an extracellular matrix protein, is expressed in injured tissues, such as the heart with myocardial infarction, and promotes angiogenesis and tissue repair. However, the molecular mechanism associated with periostin-stimulated angiogenesis and tissue repair is still unclear. In order to
Endocytosis and nuclear trafficking of adeno-associated virus type 2 are controlled by rac1 and phosphatidylinositol-3 kinase activation.
Sanlioglu, S; Benson, PK; Yang, J; Atkinson, EM; Reynolds, T; Engelhardt, JF
Journal of virology null
The ability of integrin alpha(v)beta(3) To function as a receptor for foot-and-mouth disease virus is not dependent on the presence of complete subunit cytoplasmic domains.
Neff, S; Baxt, B
Journal of virology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service