LSKMAGS15
PureProteome Magnetic Stand, 15 mL
The PureProteome Magnetic Stand, 15 mL is designed for use with PureProteome Magnetic Beads in affinity purifications (e. g., His-tag purifications or immunoprecipitations).
Synonym(s):
Magnetic Stand for 15 mL Tubes, Magnetic Stand for PureProteome Magnetic Beads
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116133
eCl@ss:
32011202
NACRES:
NA.56
Recommended Products
material
self-standing
feature
binder
manufacturer/tradename
PureProteome
technique(s)
RNA purification: suitable (with magnetic beads)
protein purification: suitable
shipped in
ambient
General description
PureProteome Magnetic Stand significantly enhance the capture efficiency of PureProteome beads, which possess a high binding capacity.
Application
PureProteome Magnetic Stand, 15 mL I suitable for:
- purification with magnetic beads
- protein purification
- in the purification of the heat shock protein 90 (Hsp90)/ Cdc37 (cell division cycle 37)/ Cyclin-dependent kinase 4 (Cdk4) complex to incubate and wash the beads with 10 bed volumes of lysis buffer
Other Notes
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protein expression and purification of the Hsp90-Cdc37-Cdk4 kinase complex from Saccharomyces cerevisiae
Verba KA and Agard DA
Bio-protocol, 7(19), e2563-e2563 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service