MAB1691
Anti-Troponin I Antibody, a.a. 186-192, clone C5
clone C5, Chemicon®, from mouse
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
clone
C5, monoclonal
species reactivity
frog, mouse, chicken, fish, human, bovine, rabbit
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
western blot: suitable
isotype
IgG2b
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Specificity
Troponin I. Reacts with Troponin I from heart and skeletal muscle.
Immunogen
Bovine cardiac Troponin I.
Epitope: a.a. 186-192
Application
Detect Troponin I using this Anti-Troponin I Antibody, clone C5 validated for use in ELISA & WB.
Immunohistochemistry. The antibody is not recommended for use on parafin sections.
Troponin sandwich ELISA (for labeling) with MAB3438.
Western blotting. Recognizes a band at approxinmately 22.5 kDa corresponding to troponin I.
Troponin I purification
Optimal working dilutions must be determined by the end user.
Troponin sandwich ELISA (for labeling) with MAB3438.
Western blotting. Recognizes a band at approxinmately 22.5 kDa corresponding to troponin I.
Troponin I purification
Optimal working dilutions must be determined by the end user.
Research Category
Metabolism
Metabolism
Research Sub Category
Muscle Physiology
Muscle Physiology
Physical form
Format: Purified
Purified immunoglobulin from Protein A Sepharose. Liquid in PBS pH 7.4 with 0.1% sodium azide as a preservative.
Storage and Stability
Maintain refrigerated at +2-8°C for up to 6 months.
Analysis Note
Control
POSITIVE CONTROL: heart or skeletal muscle.
POSITIVE CONTROL: heart or skeletal muscle.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
recommended
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Human neonatal cardiovascular progenitors: unlocking the secret to regenerative ability.
Fuentes, TI; Appleby, N; Tsay, E; Martinez, JJ; Bailey, L; Hasaniya, N; Kearns-Jonker, M
Testing null
Desmin-related cardiomyopathy in transgenic mice: a cardiac amyloidosis.
Sanbe, A; Osinska, H; Saffitz, JE; Glabe, CG; Kayed, R; Maloyan, A; Robbins, J
Proceedings of the National Academy of Sciences of the USA null
Cardioprotective effect of nicorandil, a mitochondrial ATP-sensitive potassium channel opener, prolongs survival in HSPB5 R120G transgenic mice.
Sanbe, A; Marunouchi, T; Yamauchi, J; Tanonaka, K; Nishigori, H; Tanoue, A
Testing null
Matrix identity and tractional forces influence indirect cardiac reprogramming.
Kong, YP; Carrion, B; Singh, RK; Putnam, AJ
Scientific Reports null
pH-responsive titratable inotropic performance of histidine-modified cardiac troponin I.
Palpant, NJ; Houang, EM; Sham, YY; Metzger, JM
Biophysical Journal null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service