M000037AS
Spare Ring for 37mm Aerosol Monitor Cassette
for use with 37mm Aerosol Monitor Cassette
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116116
NACRES:
NB.24
Recommended Products
material
polystyrene
Quality Level
sterility
non-sterile
filter diam.
37 mm
application(s)
environmental
compatibility
for use with 37mm Aerosol Monitor Cassette
General description
Spare Ring for Aerosol Monitor Cassette. For use with 37mm aerosol analysis monitors. Millipore′s 37mm Aerosol Analysis Monitors are transparent, disposable, plastic filter holders for use with Type MF (mixed cellulose esters) Millipore filters. A thin cellulose support seals the filter between monitor sections to distribute sample flow evenly over the filter surface. A spare ring can be inserted into the monitor cassette to increase number of stages in the aerosol monitor. This extended length be helpful or needed for different regulations.
Application
Aerosol analysis monitors are available with 0.45 μm and 0.8 μm cellulose ester membranes. Matched weight monitors contain two filters matched in weight to within 0.1mg, eliminating test filter pre-weighing in gravimetric analysis. Add spare ring to existing cassette to increase number of stages in monitor
Principle
Vacuum
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service