HTS182M
ChemiSCREEN Membrane Preparation Recombinant Human PK 2 Prokineticin Receptor
About This Item
biological source
human
Quality Level
recombinant
expressed in Chem-1 cells
form
liquid
manufacturer/tradename
ChemiScreen
Chemicon®
technique(s)
ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable
NCBI accession no.
UniProt accession no.
General description
Application
Biochem/physiol Actions
- GPCR Class: A
- Protein Target: PK2/PKR2
- Target Sub-Family: Prokineticin
Specifications
Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C.
Radioligand: [125I] MIT-1 (Perkin Elmer#: NEX-410)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.
Physical form
Liquid in packaging buffer: 50 m
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service