Skip to Content
Merck
All Photos(1)

Key Documents

HTS010RTA

Sigma-Aldrich

Ready-to-Assay CCR5 Chemokine Family Receptor Frozen Cells

rhesus macaque

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

Pricing and availability is not currently available.

Product Name

Ready-to-Assay CCR5 Chemokine Family Receptor Frozen Cells,

biological source

rhesus macaque

Quality Level

manufacturer/tradename

Ready-to-Assay

technique(s)

calcium flux assay: suitable

NCBI accession no.

detection method

fluorometric

General description

Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

CCR5 is the receptor for CC chemokines MIP-1α, MIP-1β, and RANTES (Raport et al., 1996), and is preferentially expressed on Th1 lymphocytes (Loetscher et al., 1998). CCR5 is a coreceptor for macrophage-tropic HIV, and its ligands potently inhibit HIV replication in human leukocytes (Cocchi et al., 1995). In addition, HIV-infected patients with the nonfunctional CCR5Δ32 allele exhibit delayed onset of AIDS symptoms (Samson et al., 1996), and pharmacological antagonism of CCR5 inhibits HIV-1 infection (Strizki et al., 2001). Preclinical testing of small molecule antagonists of CCR5 has been hampered by low affinity of the compounds to rodent and dog CCR5, but two such compounds, maraviroc and AD101, have been shown to have potent antagonist activity at rhesus macaque CCR5, which differs from human CCR5 by 8 amino acids (Napier et al., 2005; Billick et al., 2004). One antagonist of human CCR5, SCH-C, does not block HIV entry through rhesus macaque CCR5, and one amino acid difference is responsible for the functional difference (Billick et al., 2004). Millipore′s cloned rhesus macaque CCR5-expressing cell line is made in the Chem-1 host, which supports high levels of recombinant CCR5 expression on the cell surface and contains high levels of the promiscuous G protein Gα15 to couple the receptor to the calcium signaling pathway. Thus, the cell line is an ideal tool for screening for agonists, antagonists and modulators at CCR5.

Cell Line Description

  • GPCR Cell Lines
  • Host cells: Chem-1

Application

Rhesus Macaque CCR5 GPCR frozen cells for Calcium Flux FLIPR Assays.

Biochem/physiol Actions

  • GPCR Class: A
  • Protein Target: CCR5
  • Target Sub-Family: Chemokine

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.

Disclaimer


This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service