Skip to Content
Merck
All Photos(1)

Key Documents

EZGLP2

Millipore

Rat/Human GLP-2 ELISA Kit

detects GLP-2 in 50 μL serum, plasma or cell culture supernatent

Synonym(s):

Glucagon-like peptide-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

GLP-2 ELISA, This GLP-2 ELISA is used to measure & quantify GLP-2 levels in Metabolism & Endocrine research.

Quality Level

species reactivity

human, rat

packaging

kit of 1 × 96 wells

parameter

50 μL sample volume (4hr assay)

assay range

accuracy: 75-97%
(Human)

accuracy: 94-109%
(Rat)

sensitivity: 1 ng/mL
(50 μl sample size)

standard curve range: 1-64 ng/mL
inter-assay cv: <12%
intra-assay cv: <10%

technique(s)

ELISA: suitable

input

sample type plasma (K2 EDTA)
sample type serum

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

General description

The glucagon-like peptides GLP-1 and GLP-2 are important gut hormones released from the small intestine upon nutrient ingestion. In circulation, GLP-2 is present in two molecular forms, GLP-2 (1-33) and GLP-2 (3-33). Similar to GLP-1, active form GLP-2 (1-33) is cleaved to inactive form GLP-2 (3-33) by protease DPPIV. GLP-2 has been shown to play important roles in the regulation of gastrointestinal functions (digestion, absorption, motility, epithelial growth, and blood flow) and bone resorption. Thus, GLP-2 or its analogs may have therapeutic potentials for the treatment of diseases such as Crohn’s disease and osteoporosis.

Specificity

Human GLP-2 1-33 100%
Human GLP-2 3-33 112%
Rat GLP-2 101%

Application

Research Category
Metabolism
Research Sub Category
Osteoporosis
This assay requires 50 µl of rat or human samples and is performed at room temperature as a 4 hour assay. GLP-2 has also been detected in guinea pig, horse, porcine, and non-human primate samples.
Used to detect/quantify: GLP-2

Other Notes

Please contact Technical Service for linearity of dilution.

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is the kit compatible with cell/tissue culture supernatants that contain a DPP-IV inhibitor, considering that the same sample will be used to measure GLP-1 active?

    1 answer
    1. Yes, the kit is compatible with cell/tissue culture supernatant samples. In this case, the user would omit the Matrix Solution and replace it with their blank culture media in the background, standard, and QC wells. While the samples can contain a DPP-IV inhibitor, it is not a requirement. It is recommended to follow the same sample conditions throughout the study.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service