Skip to Content
Merck
All Photos(1)

Key Documents

EZGLPHS

Millipore

Multi Species GLP-1 ELISA Kit

measures active GLP-1 levels in 50 μL plasma

Synonym(s):

GLP-1 Active ELISA Kit, GLP-1 ELISA kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84
Pricing and availability is not currently available.

Product Name

High Sensitivity GLP-1 Active ELISA Kit, Chemiluminescent,

species reactivity

mouse, rat, human

Quality Level

packaging

kit of 1 × 96 wells

parameter

50 μL sample volume (4hr assay)

technique(s)

ELISA: suitable

input

sample type plasma (K2 EDTA)
sample type serum

application(s)

research use

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 0.14 pM
standard curve range: 0.14 -100 pM
accuracy: 96%

detection method

chemiluminescent (425nm)

shipped in

wet ice

storage temp.

2-8°C

General description

This high sensitivity GLP-1 Active ELISA kit is used for the quantification of GLP-1 (7-36) in serum, plasma and cell culture samples. The GLP-1 sequence is highly conserved between the species, with no sequence variation occurring in mammals. One kit is sufficient to measure 38 unknown samples in duplicate.

Specificity

GLP-1 (7-36) 100%
GLP-1 (7-37) 72%
GLP-1 (1-36) <3%
GLP-1 (1-37) <2%

Used to detect/quantify: GLP-1 (Active)

Application

This High Sensitivity GLP-1 Active ELISA Kit, Chemiluminescent is used to measure & quantify GLP-1 (Active) levels in Metabolism & Neuroscience research.

Research Category: Metabolism

Research Sub-Category: Obesity, Metabolic Disorders

Other Notes

Contact Technical Service for this information

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Does this work with mouse plasma? If so, how many ul of plasma will I need? Thanks

    1 answer
    1. The GLP-1 sequence is highly conserved among mammals, allowing the GLP-1 ELISA to be used with mouse samples. The sample volume is 50 ul per well and it is recommended to run samples in duplicate.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service