Skip to Content
Merck
All Photos(1)

Key Documents

CC077

Sigma-Aldrich

Human Collagen Type V

from human placenta, liquid, 1 mg/mL, suitable for cell culture, used for gel formation

Synonym(s):

Collagen Type V

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

product name

Human Collagen Type V,

biological source

human

Quality Level

Assay

95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

cell culture | mammalian: suitable

impurities

<0.5% non-collagen proteins
<1% collagen type III
<2% collagen type I
<2% collagen type IV

input

sample type mesenchymal stem cell(s)
sample type induced pluripotent stem cell(s)
sample type pancreatic stem cell(s)
sample type epithelial cells
sample type neural stem cell(s)
sample type hematopoietic stem cell(s)
sample type: human embryonic stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

General description

Collagen type V is fibril-forming collagen that is located in the lung, bone, and fetal membranes along with type I collagen. It is minor collagen present in the extracellular matrix.

Application

Human Collagen Type V has been used to study the interaction of high endothelial venule protein (hevin) with human collagen V by fluorescence polarization (FP) assay and surface plasmon resonance(SPR). It has also been used to incubate the 96 well plate to determine the protein levels in media and cell lysates of fibroblasts.

Biochem/physiol Actions

Collagen type V acts as a regulator of collagen fibrillogenesis in the cornea and skin dermis. It interacts with matrix collagens and structural proteins thereby contributing to the structural integrity of tissues. Lower levels of type V collagen lead to loss of corneal transparency and Ehler Danlos syndrome.

Physical form

Liquid, in 0.1M acetic acid, pH 3.0. No preservatives added.

Preparation Note

Purified by serial salt precipitations of a pepsin extraction of human fetal membranes and chromatography on DEAE-cellulose.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Do not thaw and refreeze.

Other Notes

Molecular composition: α1(V)]2 α2(V), native triple helix. Purity and retention of the native helical structure were monitored by SDS-PAGE, ORD measurement, and by reaction with anti-collagen type-specific monoclonal antibodies. Product Source: Human placenta, negative for HBsAg and HIV antibodies.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Gelse et al.
Advanced drug delivery reviews, 55(12), 1531-1546 (2003-11-19)
The extracellular matrix represents a complex alloy of variable members of diverse protein families defining structural integrity and various physiological functions. The most abundant family is the collagens with more than 20 different collagen types identified so far. Collagens are
Neena Philips et al.
Connective tissue research, 53(5), 373-378 (2012-02-14)
Skin aging is associated with the loss of the structural collagens and the elastin fiber components that form the extracellular matrix (ECM). It is associated with reduced transforming growth factor-β (TGF-β), angiogenesis and increased oxidative stress. Copper has been incorporated
Type V collagen in health, disease, and fibrosis
The Anatomical Record, 299(5), 613-629 (2016)
The effects of tissue pretreatment and pepsin levels on the isolation of collagens from human placenta.
Klasson, S C, et al.
Collagen and Related Research, 6, 397-408 (1986)
Shanghua Fan et al.
Structure (London, England : 1993), 29(7), 664-678 (2021-02-04)
Hevin is secreted by astrocytes and its synaptogenic effects are antagonized by the related protein, SPARC. Hevin stabilizes neurexin-neuroligin transsynaptic bridges in vivo. A third protein, membrane-tethered MDGA, blocks these bridges. Here, we reveal the molecular underpinnings of a regulatory network

Protocols

This page covers the ECM coating protocols developed for four types of ECMs on Millicell®-CM inserts, Collagen Type 1, Fibronectin, Laminin, and Matrigel.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service