Skip to Content
Merck
All Photos(1)

Key Documents

AP180P

Sigma-Aldrich

Donkey Anti-Goat IgG Antibody, HRP conjugate, Species Adsorbed

Chemicon®, from donkey

Synonym(s):

Anti-Rabbit IgG HRP, HRP Donkey Anti-Rabbit IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

goat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defense against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Application

Donkey anti-Goat IgG Antibody, HRP conjugate, Species Adsorbed is an antibody against Donkey Goat IgG for use in ELISA, IH & WB.
Immunohistochemistry:
1:500-1:5,000 dilution can be used.

Optimal working dilutions must be determined by end user.

Quality

Routinely tested on Enzyme-linked Immunosorbent Assay.

ELISA and Western Blots:
1:5,000-1:100,000 dilution can be used.

Physical form

Purified donkey secondary antibody IgG in lyophilized buffer 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 with 15 mg/mL BSA.
Reconstitution:
Reconstitute with 500 μL of sterile distilled water.

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months.
After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.
WARNING: Use of sodium azide as a preservative will substantially inhibit the enzyme activity of HRP.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Zhou et al.
Anatomical record (Hoboken, N.J. : 2007), 294(4), 729-737 (2011-02-18)
ATP sensitive K(+) (K(ATP) ) channels are important linkage of cell membrane excitability to its cellular bioenergetic state. These channels are composed of pore-forming subunits and regulatory subunits. The present study focused on the cellular expressions and localizations of these
Maria Cernea et al.
Journal of molecular endocrinology, 56(1), 39-46 (2015-10-23)
Adipose tissue expansion, resulting from adipocyte hyperplasia and/or hypertrophy, is a hallmark of obesity. Adipocytes are derived from mesenchymal stem cells (MSCs) through adipogenesis, a process involving three key steps: proliferation, commitment and differentiation. Although studies have elaborated on the
Localization of ATP-sensitive K+ channel subunits in rat submandibular gland.
Zhou, M; He, HJ; Hirano, M; Sekiguchi, M; Tanaka, O; Kawahara, K; Abe, H
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society null
Effects of corticosteroids in the development of limb muscle weakness in a porcine intensive care unit model.
Aare, S; Radell, P; Eriksson, LI; Akkad, H; Chen, YW; Hoffman, EP; Larsson, L
Physiological Genomics null
Shane A Liddelow et al.
PloS one, 7(3), e33554-e33554 (2012-03-30)
Exchange mechanisms across the blood-cerebrospinal fluid (CSF) barrier in the choroid plexuses within the cerebral ventricles control access of molecules to the central nervous system, especially in early development when the brain is poorly vascularised. However, little is known about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service