Skip to Content
Merck
All Photos(3)

Key Documents

ABE1327M

Sigma-Aldrich

Anti-DPF2

from rabbit

Synonym(s):

Zinc finger protein ubi-d4, Apoptosis response zinc finger protein, BRG1-associated factor 45D, BAF45D, D4 zinc and double PHD fingers family 2, Protein requiem

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

Related Categories

General description

Zinc finger protein ubi-d4 (UniProt: Q92785; also known as Apoptosis response zinc finger protein, BRG1-associated factor 45D, BAF45D, D4, zinc and double PHD fingers family 2, Protein requiem, DFP2) is encoded by the DPF2 (also known as BAF45D, REQ, UBID4) gene (Gene ID: 5977) in human. BAF45D is a member of the requiem/DPF protein family and is ubiquitously expressed. It may serve as a transcription factor required for the apoptosis response following survival factor withdrawal from myeloid cells. It is also believed to have a role in the development and maturation of lymphoid cells. It contains 3 zinc-finger domains (C2H2-type aa 209-232; PHD type I aa 270-330; and PHD type 2 aa 327-377). Two isoforms of BAF45D have been described that are generated via alternative splicing.

Specificity

This rabbit polyclonal antibody detects BRG1-associated factor 45D in human, mouse, and rat species. It targets an epitope within 17 amino acids from the N-terminal half. The immunogen sequence is conserved in both isoforms.

Immunogen

KLH-conjugated liner peptide that corresponding to 17 amino acids from the N-terminal region of human BRG1-associated factor 45D (DPF2).

Application

Anti-DPF2, Cat. No. ABE1327, is a highly specific rabbit polyclonal antibody that targets BRG1-associated factor 45D and has been tested in Immunocytochemistry and Western Blotting.
Western Blotting Analysis: 2 µg/mL from a representative lot detected DPF2 in 10 µg of PC12 cell lysate.

Immunocytochemistry Analysis: A 1:100-500 dilution from a representative lot detected DPF2 in A431 and HepG2 cells.

Quality

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected DPF2 in 10 µg of NIH3T3 cell lysate.

Target description

~44 kDa observed; 44.15 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service