Skip to Content
Merck
All Photos(1)

Documents

AB1254

Sigma-Aldrich

Anti-Cytochrome P450 Enzyme CYP3A4 Antibody

serum, Chemicon®

Synonym(s):

Anti-Anti-CP33, Anti-Anti-CP34, Anti-Anti-CYP3A, Anti-Anti-CYP3A3, Anti-Anti-CYPIIIA3, Anti-Anti-CYPIIIA4, Anti-Anti-HLP, Anti-Anti-NF-25, Anti-Anti-P450C3, Anti-Anti-P450PCN1, Anti-Anti-VDDR3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, pig

species reactivity (predicted by homology)

mammals, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CYP3A4(1576)

Specificity

The rabbit polyclonal serum detects CYP3A4 in humans and rats; by sequence homology of the immunogen used, cross reactivity to CYP3A7, 3A6v4;3A2v1, 3A8, 3A9 may be expected but is untested at this time.

Immunogen

A synthetic peptide containing the C-terminal sequence of TVSGA from human CYP3A4. The peptide was coupled to KLH.

Application

Anti-Cytochrome P450 Enzyme CYP3A4 Antibody detects level of Cytochrome P450 Enzyme CYP3A4 & has been published & validated for use in IH & WB.
Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry
Western blot: 1:1000-1:2000: Using microsomal membrane fractions, detects a band at ~54kDa; other bands may be present as well. Antibody has been successfully used to detect CYP3A4 on LICOR fluorescent westerns as well.

Immunohistochemistry: 10% formalin fixed, paraffin embedded tissues 1:200-1:400, microwave-citric acid antigen recovery is suggested.

Optimal working dilutions must be determined by end user.

Target description

57 kDa

Physical form

Rabbit antiSerum. Liquid containing no preservative.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Liver tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shivani Ruparel et al.
Pain, 154(11), 2363-2371 (2013-07-23)
Endogenous TRPV1 agonists such as oxidized linoleic acid metabolites (OLAMs) and the enzymes releasing them [eg, cytochrome P450 (CYP)] are up-regulated after inflammation in the rat. However, it is not known whether such agonists are elevated in human inflammatory pain
Up-regulating CYP3A4 expression in C3A cells by transfection with a novel chimeric regulator of hPXR-p53-AD.
Chen, F; Rao, XH; Yang, JL; Pan, MX; Gao, Y; Li, ZL; Li, Y; Zhu, YF; Wang, Y
Testing null
Manuel Vlach et al.
Cells, 11(23) (2022-12-12)
The goal of this study was to establish a procedure for gene delivery mediated by cationic liposomes in quiescent differentiated HepaRG™ human hepatoma cells. We first identified several cationic lipids promoting efficient gene transfer with low toxicity in actively dividing
Ulrik Kræmer Sundekilde et al.
Toxicology, 469, 153121-153121 (2022-02-11)
The circadian rhythm has profound effect on the body, exerting effects on diverse events like sleep-wake patterns, eating behavior and hepatic detoxification. The cytochrome p450 s (Cyps) is the main group of enzymes responsible for detoxification. However, the underlying mechanisms
Els Van Peer et al.
Basic & clinical pharmacology & toxicology, 114(5), 387-394 (2013-11-15)
Despite the increasing use of the minipig as a non-rodent species in general and juvenile toxicity studies, knowledge on their biotransformation processes and their ontogeny is scarce. Such data are prerequisite for the correct interpretation of non-clinical studies in this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service