Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.55134

Sigma-Aldrich

Ramage Amide AM resin

Novabiochem®

Synonym(s):

2-{[(R,S)-5-(9-Fluorenylmethyloxycarbonyl-amino)-10,11-dihydro-5H-dibenzo[a,d]cycloheptene-2-yl]oxy}acetyl-AM resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

Ramage Amide AM resin comprises Ramage′s dibenzocycloheptadienelinker [1] attached to aminomethylated polystyrene. Following Fmoc removal, the resin can be acylated under standard conditions and used in Fmoc SPPS. The linker is considerably more acid sensitive than the Rink amide or PAL linkers. This enables peptide amides to be released from the resin with 3% TFA in DCM and thus makes it a useful tool for the synthesis of acid sensitive peptides or protected peptide fragments.


Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] R. Ramage, et al. (1993) Tetrahedron Lett., 34, 6599.

Analysis Note

Colour (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.40 - 0.80 mmol/g
Swelling Volume (in DMF): lot specific result

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Novabiochem® offers a wide range of linkers and derivatized resins for Fmoc solid-phase peptide synthesis with specialized protocols.

Protocols

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Review various resins like Merrifield, trityl-based, and hydroxymethyl-functionalized for peptide immobilization for diverse applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service