8.14996
2,2-Bis(hydroxymethyl)propionic acid
for synthesis
Synonym(s):
2,2-Bis(hydroxymethyl)propionic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H10O4
CAS Number:
Molecular Weight:
134.13
MDL number:
UNSPSC Code:
12352106
EC Index Number:
225-306-3
NACRES:
NA.22
Recommended Products
Quality Level
form
powder
autoignition temp.
>300 °C
potency
>2000 mg/kg LD50, oral (Rat)
pH
2.6 (20 °C, 50 g/L in H2O)
mp
188-191 °C
transition temp
flash point >150 °C
storage temp.
2-30°C
SMILES string
OCC(CO)(C)C(=O)O
InChI
1S/C5H10O4/c1-5(2-6,3-7)4(8)9/h6-7H,2-3H2,1H3,(H,8,9)
InChI key
PTBDIHRZYDMNKB-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service