Skip to Content
Merck
All Photos(1)

Key Documents

8.14414

Sigma-Aldrich

Triethoxyoctylsilane

for synthesis

Synonym(s):

Triethoxyoctylsilane, n-Octyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H32O3Si
CAS Number:
Molecular Weight:
276.49
MDL number:
UNSPSC Code:
12352103
EC Index Number:
220-941-2
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

Assay

≥96.5% (GC)

form

liquid

potency

5110 mg/kg LD50, oral (Rat)
5142 mg/kg LD50, skin (Rabbit)

bp

98-99 °C/2.7 hPa

mp

-75.5 °C

transition temp

flash point 109 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](OCC)(OCC)(OCC)CCCCCCCC

InChI

1S/C14H32O3Si/c1-5-9-10-11-12-13-14-18(15-6-2,16-7-3)17-8-4/h5-14H2,1-4H3

InChI key

MSRJTTSHWYDFIU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.877 - 0.879
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service