Skip to Content
Merck
All Photos(1)

Key Documents

8.04808

Sigma-Aldrich

Isophthaloyl dichloride

for synthesis

Synonym(s):

Isophthaloyl dichloride, Isophthaloyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(COCl)2
CAS Number:
Molecular Weight:
203.02
MDL number:
UNSPSC Code:
12352106
EC Index Number:
202-774-7
NACRES:
NA.22

vapor pressure

0.04 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

potency

2200 mg/kg LD50, oral (Rat)
1410 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5-8.9 % (v/v)

mp

42-45 °C

transition temp

flash point 180 °C

density

1.39 g/cm3 at 17 °C

storage temp.

2-30°C

SMILES string

ClC(=O)c1cc(ccc1)C(=O)Cl

InChI

1S/C8H4Cl2O2/c9-7(11)5-2-1-3-6(4-5)8(10)12/h1-4H

InChI key

FDQSRULYDNDXQB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 43 °C
Melting range (upper value): ≤ 46 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

356.0 °F - closed cup

Flash Point(C)

180 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service