Skip to Content
Merck
All Photos(1)

Key Documents

8.00251

Sigma-Aldrich

Triethyl citrate

for synthesis

Synonym(s):

Triethyl citrate, Citric acid triethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOC2H5)(CH2COOC2H5)2
CAS Number:
Molecular Weight:
276.28
MDL number:
UNSPSC Code:
12352108
EC Index Number:
201-070-7
NACRES:
NA.22

vapor pressure

0.0025 hPa ( 25 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

potency

>3200 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

294 °C/1013 hPa

mp

-55 °C

transition temp

flash point 150 °C

solubility

65 g/L

density

1.135 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

O(CC)C(=O)C(O)(CC(=O)OCC)CC(=O)OCC

InChI

1S/C12H20O7/c1-4-17-9(13)7-12(16,11(15)19-6-3)8-10(14)18-5-2/h16H,4-8H2,1-3H3

InChI key

DOOTYTYQINUNNV-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 98 %
Density (d 20 °C/ 4 °C): 1.138 - 1.141
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service