Skip to Content
Merck
All Photos(1)

Key Documents

W308307

Sigma-Aldrich

Triethyl citrate

≥99%, FCC, FG

Synonym(s):

Ethyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOC2H5)(CH2COOC2H5)2
CAS Number:
Molecular Weight:
276.28
FEMA Number:
3083
Beilstein:
1801199
EC Number:
Council of Europe no.:
11762
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.512
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 117
FDA 21 CFR 175.300
FDA 21 CFR 175.320
FDA 21 CFR 181.27
FDA 21 CFR 184.1911

vapor density

9.7 (vs air)

vapor pressure

1 mmHg ( 107 °C)

Assay

≥99%

refractive index

n20/D 1.442 (lit.)

bp

235 °C/150 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

InChI

1S/C12H20O7/c1-4-17-9(13)7-12(16,11(15)19-6-3)8-10(14)18-5-2/h16H,4-8H2,1-3H3

InChI key

DOOTYTYQINUNNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethyl citrate can be used as a plasticizer and sequestrant in the food industry.

Application


  • Cosmetically Approved Short-Chain Alcohol/Triethyl Citrate/Water Surfactant-Free Microemulsions and Potential Application to Transdermal Penetration of α-Arbutin.: This study investigates the formulation of surfactant-free microemulsions using triethyl citrate, exploring its potential in enhancing the transdermal penetration of α-arbutin. The results suggest significant improvement in skin delivery, highlighting the cosmetic and pharmaceutical potential of triethyl citrate (Zhang et al., 2024).

  • Understanding the Interaction of Thermal, Rheological, and Mechanical Parameters Critical for the Processability of Polyvinyl Alcohol-Based Systems during Hot Melt Extrusion.: This research delves into the role of triethyl citrate as a plasticizer in polyvinyl alcohol-based systems, essential for optimizing hot melt extrusion processes. The findings emphasize the importance of precise thermal and mechanical control in pharmaceutical applications (Hess et al., 2024).

  • The effect of additives on the biodegradation of polyhydroxyalkanoate (PHA) in marine field trials.: Triethyl citrate is evaluated as an additive to improve the biodegradation of polyhydroxyalkanoates in marine environments. This study presents its efficacy in enhancing environmental sustainability through accelerated degradation (Read et al., 2024).

  • In-situ forming PLGA implants: Towards less toxic solvents.: Investigating the use of triethyl citrate as a less toxic solvent alternative in PLGA implants, this paper demonstrates its potential in reducing toxicity while maintaining implant efficacy, beneficial for pharmaceutical applications (Ramos et al., 2024).

  • Influence of Low-Molecular-Weight Esters on Melt Spinning and Structure of Poly(lactic acid) Fibers.: This article explores the impact of triethyl citrate on the properties of poly(lactic acid) fibers, highlighting its role in enhancing fiber structure and processability for industrial applications (Gzyra-Jagieła et al., 2024).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictionary of Food ingredients, 144-144 (2013)
Food Additives, 682-682 (1999)
Gavin P Andrews et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 69(1), 264-273 (2008-01-01)
The aim of this highly novel study was to use hot-melt extrusion technology as an alternative process to enteric coating. In so doing, oral dosage forms displaying enteric properties may be produced in a continuous, rapid process, providing significant advantages
Kakuyou Ogawa et al.
Biological & pharmaceutical bulletin, 37(9), 1559-1563 (2014-09-02)
Zerumbone 1 is an 11-membered cyclic sesquiterpene obtained from the rhizomes of Zingiber zerumbet SMITH (Zingiberaceae). In this study, we investigated the structure-activity relationship of 1, α-humulene (2), tetrahydrozerumbone stereoisomers (3-5), and tetrahydrozerumbone derivatives (6-9). The oxygen-containing functional groups and
S Monfort et al.
International journal of food microbiology, 145(2-3), 476-482 (2011-02-19)
This paper is an evaluation of the lethal effectiveness of a successive application of pulsed electric fields (PEFs) and heat treatment in liquid whole egg (LWE) in the presence of different additives on the population of Salmonella serovar Enteritidis. Synergistic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service