Please see the composition for each of the 3 buffers included in this kit:
Ni-NTA Bind Buffer: 50 mM NaH2PO4, pH 8.0; 300 mM NaCl; 10 mM imidazole
Ni-NTA Wash Buffer: 50 mM NaH2PO4, pH 8.0; 300 mM NaCl; 20 mM imidazole
Ni-NTA Elute Buffer: 50 mM NaH2PO4, pH 8.0; 300 mM NaCl; 250 mM imidazole
70899
Ni-NTA Buffer Kit
Synonym(s):
Buffer Kit for Ni-NTA, Ni-NTA Buffer
About This Item
Recommended Products
manufacturer/tradename
Novagen®
Quality Level
storage condition
OK to freeze
shipped in
ambient
storage temp.
10-30°C
General description
Components
•125 ml4X Ni-NTA Wash Buffer
•50 ml4X Ni-NTA Elute Buffer
Warning
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Repr. 1B - Skin Corr. 1C
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
YeastBuster™ Protein Extraction Reagent for Fast, Efficient Extraction of Proteins from Yeast
-
Hi I want to buy this kit: 70899 Ni-NTA Buffer Kit and want to also know the buffer composition. Since I am doing native protein purification its important for me to know the composition of the buffers. Please inform where I can find it. Thanks
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service