Skip to Content
Merck
All Photos(1)

Key Documents

71456

Millipore

BugBuster® Master Mix

Synonym(s):

BugBuster® Protein Extraction Reagent with Benzonase® Nuclease and rLysozyme solution, Cell Lysis Master Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106511
NACRES:
NA.32
Pricing and availability is not currently available.

form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

2-8°C

General description

The BugBuster® Master Mix enables efficient protein purification and other related applications. It replaces the need for mechanical methods like the French press or sonication. The two available package sizes contain sufficient reagents for extracting proteins from either 20 grams or 100 grams of cell paste.

Application

BugBuster® Master Mix has been used:
  • to resuspend and lyse the cell pellets during protein purification[1]
  • to extract proteins for immunoblotting[2]
  • as a bacterial lysis buffer for the preparation of inclusion bodies[3]


    

Biochem/physiol Actions

BugBuster® Protein Extraction Reagent is specifically designed to gently disrupt the cell wall of E. coli, allowing the release of soluble proteins. This unique formulation utilizes a detergent mix that can effectively perforate the cell wall without denaturing the soluble proteins. By combining BugBuster Reagent, rLysozyme solution, and Benzonase Nuclease, our BugBuster Master Mix enhances protein extraction efficiency and simplifies downstream processing of protein extracts.

Features and Benefits

  • Efficient lysis of Gram-positive and Gram-negative bacteria
  • Maximum recovery of active, soluble protein
  • Capable of cell wall perforation without denaturing soluble protein
  • Useful for the preparation of high purity inclusion bodies
  • Compatible with protease and phosphatase inhibitor cocktail

  • Offers a convenient, fast, and cost-effective solution for releasing the expressed target protein.
  • The Master Mix eliminates the need to dilute or perform separate addition steps.

Warning

Toxicity: Standard Handling (A)

Legal Information

BUGBUSTER is a registered trademark of Merck KGaA, Darmstadt, Germany
Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew Thomas Doyle et al.
Molecular cell, 81(9), 2000-2012 (2021-03-12)
The β-barrel assembly machine (BAM) integrates β-barrel proteins into the outer membrane (OM) of Gram-negative bacteria. An essential BAM subunit (BamA) catalyzes integration by promoting the formation of a hybrid-barrel intermediate state between its own β-barrel domain and that of
Development of a synthetic biosensor for chemical exchange MRI utilizing in silico optimized peptides
Fillion AJ, et al.
Nmr in Biomedicine, 36, e5007-e5007 (2023)
Yong Y Peng et al.
Bioengineered, 5(6), 378-385 (2014-12-09)
The collagen like domain Scl2 from Streptococcus pyogenes has been proposed as a potential biomedical material. It is non-cytotoxic and non-immunogenic and can be prepared in good yield in fermentation. The Scl2 collagen domain is about a quarter of the
RT-QuIC assays for prion disease detection and diagnostics
Orru CD, et al.
Journal of biological and chemical chronicles, 185-203 (2017)
Daniel G Isom et al.
Proteins, 79(4), 1034-1047 (2011-03-10)
Protein thermodynamic stability is a fundamental physical characteristic that determines biological function. Furthermore, alteration of thermodynamic stability by macromolecular interactions or biochemical modifications is a powerful tool for assessing the relationship between protein structure, stability, and biological function. High-throughput approaches

Articles

Citation Spotlight: BugBuster® Protein Extraction Reagent for Efficient Protein Extraction from Bacterial Pathogens

Related Content

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

Cell lysis and protein extraction methods overview various techniques, from detergent solubilization to mechanical disruption, supporting research needs.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service