Skip to Content
Merck
All Photos(1)

Key Documents

681675

Sigma-Aldrich

Wortmannin

≥98% (HPLC), solid, PI3-kinase inhibitor, Calbiochem

Synonym(s):

Wortmannin, KY 12420, MLCK Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24O8
CAS Number:
Molecular Weight:
428.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Wortmannin, Wortmannin, CAS 19545-26-7, is a cell-permeable, potent, selective, and irreversible inhibitor of PI3-Kinase (IC₅₀ = 5 nM). Does not affect any upstream signaling events.

description

Merck USA index - 14, 10053

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[o]1c2c3c(c1)C(=O)O[C@@H]([C@@]3(C4=C([C@H]5[C@](C[C@H]4OC(=O)C)(C(=O)CC5)C)C2=O)C)COC

InChI

1S/C23H24O8/c1-10(24)30-13-7-22(2)12(5-6-14(22)25)16-18(13)23(3)15(9-28-4)31-21(27)11-8-29-20(17(11)23)19(16)26/h8,12-13,15H,5-7,9H2,1-4H3/t12-,13+,15+,22-,23-/m0/s1

InChI key

QDLHCMPXEPAAMD-QAIWCSMKSA-N

General description

A cell-permeable, fungal metabolite that acts as a potent, selective, and irreversible inhibitor of phosphatidylinositol 3-kinase in purified preparations and cytosolic fractions (IC50 = 5 nM). Blocks the catalytic activity of PI 3-kinase without affecting the upstream signaling events. Preincubation of fibroblasts with wortmannin abolishes PDGF-mediated Ins(3,4,5)P3 formation (IC50 = 5 nM). Also blocks the metabolic effects of insulin in isolated rat adipocytes without affecting the insulin receptor tyrosine kinase activity. Inhibits MAP kinase activation induced by platelet activating factor in guinea pig neutrophils (200 - 300 nM). Also inhibits other kinases such as myosin light chain kinase (IC50 = 200 nM) and PI 4-kinase at concentrations higher than that required for inhibition of PI 3-kinase. Also blocks phospholipase D. A 10 mM (1 mg/233 µl) solution of Wortmannin (Cat. No. 681676) in DMSO is also available.
A fungal metabolite that acts as a potent, selective, cell-permeable and irreversible inhibitor of phosphatidylinositol 3-kinase (PI 3-kinase) in purified preparations and cytosolic fractions (IC50 = 5 nM). Preincubation of fibroblasts with wortmannin abolishes PDGF-mediated Ins(3,4,5)P3 formation (IC50 = 5 nM). Blocks the catalytic activity of PI 3-kinase without affecting the upstream signaling events. Also blocks the metabolic effects of insulin in isolated rat adipocytes without affecting the insulin receptor tyrosine kinase activity. Inhibits MAP kinase activation induced by platelet activating factor in guinea pig neutrophils (0.2-0.3 µM). Inhibits osteoclastic bone resorption. Inhibits other kinases, such as myosin light chain kinase (IC50 = 200 nM) and PI 4-kinase at concentrations 100-fold higher than that required for the inhibition of PI 3-kinase. Also blocks phospholipase D.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
phosphatidylinositol-3-kinase
Product does not compete with ATP.
Reversible: no
Target IC50: 5 nM against phosphatidylinositol 3-kinase; 5 nM against PDGF-mediated Ins(3,4,5)P3 formation; 200 nM against myosin light chain kinase

Packaging

Packaged under inert gas

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Cross, M.J., et al. 1995. J. Biol. Chem. 270, 25352.
Nakamura, I., et al. 1995. FEBS Lett. 361, 79.
Ferby, I.M., et al. 1994. J. Biol. Chem. 269, 30485.
Okada, T., et al. 1994. J. Biol. Chem. 269, 3568.
Wymann, M.P. and Arcaro, A. 1994. Biochem. J.298, 517.
Arcaro, A. and Wymann, M.P. 1993. Biochem. J.296, 297.
Nakanishi, S., et al. 1992. J. Biol. Chem. 267, 2157.
Bonser, R.W., et al. 1991. Br. J. Pharmacol.103, 1237.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Acute Tox. 2 Dermal

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pore-forming protein I?I?-CAT promptly responses to fasting with capacity to deliver macromolecular nutrients.
Shi, et al.
Faseb Journal, 36, e22533-e22533 (2022)
Shuichi Tsuruoka et al.
Scientific reports, 10(1), 20548-20548 (2020-11-27)
Acidosis is associated with E. coli induced pyelonephritis but whether bacterial cell wall constituents inhibit HCO3 transport in the outer medullary collecting duct from the inner stripe (OMCDi) is not known. We examined the effect of lipopolysaccharide (LPS), on HCO3 absorption
Oliver G Rössler et al.
Journal of neurochemistry, 88(5), 1240-1252 (2004-03-11)
We have investigated the molecular mechanisms of neurotrophin-mediated cell survival in HT22 cells, a murine cell line of hippocampal origin, expressing the brain-derived neurotrophic factor (BDNF) receptor TrkB as well as the TrkB.T1 splice variant. Stimulation with BDNF protected HT22-TrkB
Zhong Zhao et al.
Communications biology, 5(1), 730-730 (2022-07-23)
Maintaining water balance is a real challenge for amphibians in terrestrial environments. Our previous studies with toad Bombina maxima discovered a pore-forming protein and trefoil factor complex βγ-CAT, which is assembled under tight regulation depending on environmental cues. Here we
Jie-Ning Li et al.
Frontiers in cell and developmental biology, 9, 671244-671244 (2021-07-23)
MicroRNAs (miRNAs) are small non-coding RNAs which post-transcriptionally suppress target mRNAs expression and/or translation to modulate pathophyological processes. Expression and function of miRNAs are fine-tuned by a conserved biogenesis machinery involves two RNase-dependent processing steps of miRNA maturation and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service