567703
Spermine NONOate
Complex of nitric oxide (NO) with spermine.
Synonym(s):
Spermine NONOate, N-(2-Aminoethyl)-N-(2-hydroxy-2-nitrosohydrazino)-1,2-ethylenediamine, NOC-22
About This Item
Recommended Products
Quality Level
Assay
≥98% (UV)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
color
white
solubility
0.1 M NaOH: soluble
water: soluble
shipped in
ambient
storage temp.
−20°C
InChI
1S/C10H26N6O2/c11-5-3-8-13-7-1-2-9-15(10-4-6-12)16(18)14-17/h13,18H,1-12H2/p+3
General description
Biochem/physiol Actions
Convenient reagent for preparing aqueous solutions of NO
Packaging
Warning
Other Notes
Hrabie, J.A., et al. 1993. J. Org. Chem.58, 1472.
Maragos, C.M., et al. 1993. Cancer Res. 53, 564.
Wink, D.A., et al. 1993. Proc. Natl. Acad. Sci. USA90, 9813.
Maragos, C.M., et al. 1991. J. Med. Chem. 34, 3242.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service