Skip to Content
Merck
All Photos(2)

Key Documents

5.38099

Sigma-Aldrich

GS-493

Synonym(s):

GS-493, 4-((2 Z)-2-(1,3- bis(4-Nitrophenyl)-5-oxo-1,5-dihydro-4H-pyrazol-4-yliden)hydrazino)benzenesulfonic acid, ( Z)-4-(2-(1,3- bis(4-Nitrophenyl)-5-oxo-1H-pyrazol-4(5H)-ylidene)hydrazinyl)benzenesulfonic acid, SHP2 Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14N6O8S
Molecular Weight:
510.44
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 1 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, bioavailable, non-toxic p-nitro analog of PHPS1 (Cat. No. 540213) that exhibits enhanced potency and selectivity. Acts as a phosphotyrosine mimetic, substrate-competitive and reversible catalytic domain inhibitor of SHP2 (IC50 = 0.071, 2.08 & 3.17 µM for SHP2, SHP1 & PTP1B, respectively). Dose-dependently reverts HGF-stimulatd epithelial-mesenchymal transition of HPAF II cells and blocks human lung adenocarcinoma LXFA 526L anchorage-independent growth. Shown to significantly retard LXFA 526L xenografted tumor growth in mice (46 mg/kg/day, i.p., q.d.).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
SHP2
Reversible: yes
Target IC50: 0.071, 2.08 & 3.17 &micro

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Grosskopf, S., et al. 2015. ChemMedChem.10, 815.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service