Skip to Content
Merck
All Photos(1)

Key Documents

5295

Millipore

Polyvinylpyrrolidone

Synonym(s):

Polyvinylpyrrolidone, PVP, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.21

description

RTECS - TR8370000
Merck USA index - 14, 7697

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

water: 20%

cation traces

heavy metals: ≤10 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

N1(CCCC1=O)C=C

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

General description

Polymer used as a pharmaceutical aid, complexing agent, and a solubilizer.
Polymer used as a pharmaceutical aid, complexing agent, and solubilizer.

Application


  • Synthesis of Controllable Superparamagnetic Nano Fe(3)O(4) Based on Reduction Method for Colloidal Clusters of Magnetically Responsive Photonic Crystals.: This study discusses the synthesis of superparamagnetic nano Fe(3)O(4) using polyvinylpyrrolidone (PVP) as a stabilizing agent. The research highlights the applications of PVP in enhancing the stability and controllability of magnetic nanoparticles, which are crucial for the development of advanced photonic crystal materials (Chen et al., 2024).

  • A Low-Cost Protocol Using the Adjunctive Action of Povidone-Iodine Irrigations and Sodium Hypochlorite Rinsing Solution in Step 2 of Periodontal Therapy for Patients with Stage III-IV Periodontitis: A Single-Blind, Randomized Controlled Trial.: This clinical trial explores the use of povidone-iodine, a form of polyvinylpyrrolidone, in periodontal therapy. The findings suggest significant improvements in patient outcomes, demonstrating the effectiveness of PVP-iodine in reducing periodontal pathogens (Kardaras et al., 2024).

  • Polyvinylpyrrolidone Assisted One-Pot Synthesis of Size-Tunable Cocktail Nanodrug for Multifunctional Combat of Cancer.: This study demonstrates the role of polyvinylpyrrolidone in the synthesis of size-tunable nanodrugs for cancer treatment. The use of PVP enhances the stability and delivery efficiency of the nanodrugs, providing a promising approach for targeted cancer therapy (Wang et al., 2024).

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service