Skip to Content
Merck
All Photos(4)

Key Documents

234257

Sigma-Aldrich

Polyvinylpyrrolidone

powder, average Mw ~29,000

Synonym(s):

PVP, Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

mol wt

average Mw ~29,000

ign. residue

≤0.5%

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PVP has biocompatibility, low toxicity, adhesive characteristics, complexing stability, relatively inert behavior, and is resistant to thermal degradation. It is readily prepared by the polymerization of N-vinyl-2-pyrrolidone or by the Reppe synthesis technique.

Application

PVP is one of the constituents used in the synthesis and coating of silver nanocubes. It serves as a reducing agent[1] and colloidal stabilizer[2] in the synthesis of palladium nanobars[3][1] and nanoplates.[4] It is also used as a capping agent in the synthesis of silver nanospheres. General applications of PVP are: in pharmaceutical, food, beverage, cosmetic, toiletry and photographic industries.

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lindsey T Brinton et al.
Cancers, 12(6) (2020-06-18)
Acute myeloid leukemia (AML) is a hematopoietic stem-cell-derived leukemia with often successive derived driver mutations. Late onset acquisition of internal tandem duplication in FLT3 (FLT3-ITD) at a high variant allele frequency often contributes to full transformation to a highly proliferative
Synthesis and characterization of fivefold twinned nanorods and right bipyramids of palladium
Xiong Y, et al.
Chemical Physics Letters, 440(4), 273-278 (2007)
Yujie Xiong et al.
Journal of the American Chemical Society, 127(48), 17118-17127 (2005-12-01)
The rapid reduction of Na(2)PdCl(4) by ethylene glycol in the presence of poly(vinyl pyrrolidone) (PVP) has recently been demonstrated as a convenient method of generating Pd cubooctahedra and twinned nanoparticles. Here we describe a new procedure where Pd triangular or
Yujie Xiong et al.
Journal of the American Chemical Society, 129(12), 3665-3675 (2007-03-06)
This paper describes a simple and versatile method for growing highly anisotropic nanostructures of Pd, single-crystal nanobars bounded by {100} facets and single-crystal nanorods with their side surfaces enclosed by {100} and {110} facets. According to thermodynamic arguments, Pd atoms
Morphological Evolution of Single-Crystal Ag Nanospheres During the Galvanic Replacement Reaction with HAuCl4
Kim MH, et al.
The Journal of Physical Chemistry C, 112(21), 78772-77876 (2008)

Articles

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

See All

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service