Skip to Content
Merck
All Photos(1)

Key Documents

513022

Sigma-Aldrich

PD 150606

A cell-permeable, non-competitive, selective non-peptide calpain inhibitor [Ki = 210 nM for calpain-1 and 370 nM for calpain-2.

Synonym(s):

PD 150606, 3-(4-Iodophenyl)-2-mercapto-(Z)-2-propenoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7IO2S
CAS Number:
Molecular Weight:
306.12
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL
methanol: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, non-competitive, selective non-peptide calpain inhibitor [Ki = 210 nM for calpain-1 (Cat. Nos. 208712 and 208713) and 370 nM for calpain-2 (Cat. Nos. 208715 and 208718)] directed towards the calcium binding sites of calpain. Exhibits high specificity for calpains relative to other proteases, such as cathepsin B and cathepsin L. Does not shield calpain against inactivation by the active-site inhibitor trans-(epoxysuccinyl)-L-leucyl-amido-3-methylbutane. Reportedly prevents dexamethasone-induced apoptosis in thymocytes.
A cell-permeable, selective non-peptide calpain inhibitor (Ki = 210 nM for calpain I and 370 nM for calpain II) directed towards the calcium- binding sites of calpain. Exhibits high specificity for calpains relative to other proteases, such as cathepsin B and L. PD 150606 is a non-competitive inhibitor with respect to the substrate and does not shield calpain against inactivation by the active-site inhibitor trans-(epoxysuccinyl)-L-leucyl-amido-3-methylbutane. Reportedly prevents dexamethasone-induced apoptosis of thymocytes.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Calpain-1
Product does not compete with ATP.
Reversible: no
Target Ki: 210 nM for calpain-1 and 370 nM for calpain-2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Lin, G.D., et al. 1997. Nat. Struct. Biol. 4, 539.
Squier, M.K., and Cohen, J.J. 1997. J. Immunol. 158, 3960.
Edelstein, C.L., et al. 1996. Kidney Int. 50, 1150.
Wang, K.K.W., et al. 1996. Proc. Natl. Acad. Sci. USA 93, 6687.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehmet Şerif Aydın et al.
iScience, 26(10), 107715-107715 (2023-09-13)
Trauma, vascular events, or neurodegenerative processes can lead to axonal injury and eventual transection (axotomy). Neurons can survive axotomy, yet the underlying mechanisms are not fully understood. Excessive water entry into injured neurons poses a particular risk due to swelling

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service