Skip to Content
Merck
All Photos(1)

Key Documents

5.06029

Sigma-Aldrich

LipidGreen2

Synonym(s):

LipidGreen2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29NO4
Molecular Weight:
371.47
UNSPSC Code:
12352202
NACRES:
NA.21

Pricing and availability is not currently available.

Assay

≥97% (HPLC)

Quality Level

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable N-dimethylallylated 3-hydroxyindole based compound that is shown to selectively stain neutral lipids, but not phospholipids, in various cells lines with bright fluorescence and with minimal non-specific background. Generates stronger staining with lipid droplets (~10 µM) when compared to Nile Red. Shown to be more effective than BODIPY® 493/503 in assessing hepatotoxicity of several known compounds using HepG2 cells. Also suitable for staining fat deposits in developing zebrafish embryos. Can be used in conjunction with LipidToxt red dye for multicolor imaging of neutral lipids.
A cell-permeable N-dimethylallylated 3-hydroxyindole based compound that is shown to selectively stain neutral lipids, but not phospholipids, in various cells lines with bright fluorescence and with minimal non-specific background. Generates stronger staining with lipid droplets (~10 µM) when compared to Nile Red. Shown to be more effective than BODIPY® 493/503 in assessing hepatotoxicity of several known compounds using HepG2 cells. Also suitable for staining fat deposits in developing zebrafish embryos. Can be used in conjunction with LipidToxt red dye for multicolor imaging of neutral lipids.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Chun, H., et al, 2013, Mol Biosys.9, 630.

Legal Information

BODIPY is a registered trademark of Life Technologies
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service