Skip to Content
Merck
All Photos(1)

Key Documents

5.05985

Sigma-Aldrich

CFTR Activator, Cact-A1

Synonym(s):

CFTR Activator, Cact-A1, Cystic Fibrosis Transmembrane Conductance Regulator Activator, Cact-A1, 5-(( Z)-2-(2-(Allyloxy)phenyl)-1-cyanovinyl)-3-amino-1H-pyrazole-4-carbonitrile, ( Z)-3-(2-(2-(Allyloxy)phenyl)-1-cyanovinyl)-5-amino-1H-pyrazole-4-carbonitrile, Cystic Fibrosis Transmembrane Conductance Regulator Activator, Cact-A1, 5-((Z)-2-(2-(Allyloxy)phenyl)-1-cyanovinyl)-3-amino-1H-pyrazole-4-carbonitrile, (Z)-3-(2-(2-(Allyloxy)phenyl)-1-cyanovinyl)-5-amino-1H-pyrazole-4-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13N5O
CAS Number:
Molecular Weight:
291.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A non-cytotoxic (30 µM for 48 h in human airway epithelial Calu-3 cultures) aminopyrazolocarbonitrile compound that is shown to activate both wt- and ΔF508-CFTR channel activity in a cAMP-independent, reversible, and CFTRinh-172- (Cat. Nos. 219670 & 219674 ) blockable (IC50 & IC100 = 0.37 & 30 µM, respectively, against 30 µM Cact using Fisher rat thyroid/FRT cells with wt-CFTR) manner, without affecting CaCC (Calcium-activated chloride channel) function or cellular levels of Ca2+ and cAMP. The maximum activation inducible by Cact-A1 (ECmax = 13 to 30 µM in Cl- current induction using in Fisher rat thyroid/FRT and primary human bronchial epithelial/HBE cells) is comparable to that achievable via cAMP stimulation by Forskolin treatment (ECmax = 20 µM; Cat. Nos. 344270 & 344282), which cannot be further enhanced by Forskolin co-treatment in cells expressing wt-CFTR, although additive effects are seen when Cact-A1 and Forskoklin are applied at suboptimal concentrations to cells with wt-CFTR or when the two drugs are applied at respective ECmax to cells with surface ΔF508-CFTR. Neither Cact-A1 nor Forskolin alone (at 30 µM; and 20 µM;, respectively) can effectively activate FRT with surface G551D-CFTR or primary HNPE (human nasal polyp epithelial) cells with surface G551D/Y1092X-CFTR, while Cact-A1-Forskolin co-treatment results in synergistic activation and maximally achievable activation is reported with a three-drug co-treatment using Cact-A1, Forskolin, and VX-770 (at 30, 20, and 10 µM, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CFTR
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Namkung, W., et al. 2013. Mol. Pharmacol.84, 384.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service