Skip to Content
Merck
All Photos(1)

Key Documents

5.00508

Sigma-Aldrich

ATR Inhibitor III, ETP-46464

Synonym(s):

ATR Inhibitor III, ETP-46464, ATM Inhibitor III, mTOR Inhibitor XIII, PI 3-K Inhibitor XIX, ETP46464, 2-methyl-2-(4-(2-oxo-9-(quinolin-3-yl)-2H-[1,3]oxazino[5,4-c]quinolin-1(4H)-yl)phenyl)propanenitrile, ATM and Rad3-Related Inhibitor III, Ataxia-Telangiectasia Mutated Inhibitor III,

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H22N4O2
CAS Number:
Molecular Weight:
470.52
UNSPSC Code:
12352200

Assay

≥99% (HPLC)

Quality Level

form

powder

potency

14 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

InChI

1S/C30H22N4O2/c1-30(2,18-31)23-8-10-24(11-9-23)34-28-22(17-36-29(34)35)16-33-27-12-7-19(14-25(27)28)21-13-20-5-3-4-6-26(20)32-15-21/h3-16H,17H2,1-2H3

InChI key

DPLMXAYKJZOTKO-UHFFFAOYSA-N

General description

A cell-permeable quinoline-containing heterotricyclic compound that acts as a potent inhibitor against mTOR, ATR, DNA-PK, PI 3-Kα, and ATM (IC50 = 0.6, 14, 36, 170, and 545 nM, respectively) and preferentially suppresses radiation-induced cellular ATR activity (>90% at 500 nM) over ATM or DNA-PK activity (IC50 >5 M) in U2OS cells, while exhibiting much reduced or little potency toward a panel of 26 other kinases (>55% inhibition at 5 M). Shown to synergize with UCN-01 (Cat. No. 539644) in replicative stress induction in S-phase replicating U2OS (1.5-, 61-, and 190-fold of control RS, respectively, with 1 M ETP-46464, 50 nM UCN-01, or combined treatment).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
A cell-permeable quinoline-containing heterotricyclic compound that acts as a potent inhibitor against mTOR, ATR, DNA-PK, PI 3-Kα, and ATM activity (IC50 = 0.6, 14, 36, 170, and 545 nM, respectively, in cell-free kinase assays) and preferentially suppresses ionization radiation/IR-induced cellular ATR activity (>90% inhibition of IR-induced Chk1 pSer345 at 500 nM) over ATM or DNA-PK activity (IC50 >5 µM against IR-induced ATM pSer189, Chk2 pThr68, DNA-PKcs pSer2056, and γH2AX) in U2OS cells, while exhibiting much reduced or little potency toward a panel of 26 other kinases (>55% inhibition at 5 µM). Shown to synergize with UCN-01 (Cat. No. 539644) in replicative stress/RS induction in S-phase replicating U2OS (% population with pan-nuclear γH2AX staining = 6.0% and 0.2%, respectively, with or without 8 h 5 µM ETP-46464 treatment; 1.5-, 61-, and 190-fold of control RS, respectively, with 1 µM ETP-46464 alone, 50 nM UCN-01 alone, or combined treatment). Likewise, ETP-46464 is also demonstrated to further enhance oncogenic mutations-caused RS elevation in MEF cultures (% RS population without/with 5 µM ETP-46464 treatment = 0.3/0.6, 1.3/2.5, 0.9/3.0, 3.9/11.4, respectively, in wt, p53-/-, Cyclin E-transfected wt, and Cyclin E-transfected p53-/- MEF).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ATR
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Thanasoula, M., et al. 2012. EMBO J.31, 3398.
Toledo, L.I., et al. 2011. Nat Struct Mol Biol.18, 721.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis I Toledo et al.
Nature structural & molecular biology, 18(6), 721-727 (2011-05-10)
Oncogene activation has been shown to generate replication-born DNA damage, also known as replicative stress. The primary responder to replicative stress is not Ataxia-Telangiectasia Mutated (ATM) but rather the kinase ATM and Rad3-related (ATR). One limitation for the study of
Maria Thanasoula et al.
The EMBO journal, 31(16), 3398-3410 (2012-07-31)
Shelterin component TRF2 prevents ATM activation, while POT1 represses ATR signalling at telomeres. Here, we investigate the mechanism of G2/M arrest triggered by telomeres uncapped through TRF2 or POT1 inhibition in human cells. We find that telomere damage-activated ATR and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service