Skip to Content
Merck
All Photos(2)

Key Documents

492031

Sigma-Aldrich

NS8593

An aminobenzimidazole derivative that selectively and reversibly blocks small conductance Ca2+-activated K+ channels in a Ca2+-dependent manner.

Synonym(s):

NS8593, (R)-N-(Benzimidazol-2-yl)-1,2,3,4-tetrahydro-1-naphthylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3
CAS Number:
Molecular Weight:
263.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

C1(NC(N[C@@H]2CCCC3=C2C=CC=C3)=N4)=C4C=CC=C1

InChI key

XZIZUQSOFMLIIR-CQSZACIVSA-N

General description

An aminobenzimidazole derivative that selectively and reversibly blocks small conductance Ca2+-activated K+ channels (SK1-3; Kd = 420 nM, 600 nM, and 730 nM for SK1, SK2, SK3, respectively) in a Ca2+-dependent manner. It mediates channel gating by interacting with gating structures deep within the inner pore vestibule where Ser507 and Ala532 are deemed to be important for inhibition. Interacts at a site that is distinct from the apamin binding site in SK channels. Can access high-affinity binding sites from both the inside and outside of the cell membrane. Does not affect QT intervals, but prolongs the atrial effective refractive period and prevents acetylcholine-induced atrial fibrillations in ex vivo and in vivo models. Also shown to reversibly block TRPM7 channel (IC50 = 1.6 mM) in smooth muscle cells, primary podocytes, HEK293 cells expressing TRPM7, and ventricular myocytes in a Mg2+-dependent manner and blocks the motility of cells in culture.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chubanov, V., et al. 2012. Brit. Journ. Pharm.166, 1357.
Jenkins, D., et al. 2011. Hypertension.57, 1129.
Jenkins, D., et al. 2011. Mol. Pharm.79, 899.
Jenkins, D., et al. 2011. Mol. Pharm.79, 899.
Lasse, S., et al. 2011. J. Cardiov. Pharm.57, 672.
Diness, J., et al. 2010. Circ Arrhythm Electrophusiol.3, 380.
Ji, H., et al. 2009. Eur. J. Neurosci.9, 1883.
Strobaek, D., et al. 2006. Mol. Pharm.70, 1771.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service