Skip to Content
Merck
All Photos(1)

Key Documents

401003

Sigma-Aldrich

(±)-Ibuprofen

A nonsteroidal anti-inflammatory drug (NSAID) that acts as a reversible and competitive inhibitor of cyclooxygenase 1 (COX-1) (IC₅₀ = 4.85 µM).

Synonym(s):

(±)-Ibuprofen, [(±)-2-(4-Isobutylphenyl)-propionic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 4881

Assay

≥98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

ethanol: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

OC(=O)C(C)c1ccc(cc1)CC(C)C

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)

InChI key

HEFNNWSXXWATRW-UHFFFAOYSA-N

General description

A nonsteroidal anti-inflammatory drug (NSAID) that acts as a reversible, competitive, non-selective cyclooxygenase (COX) inhibitor (IC50 = 4.85 µM for purified COX-1 and 223 µM for purified COX-2). Also reported to inhibit COX-1 and COX-2 activity in intact bovine aortic endothelial cells (BAEC) (IC50 = 7 µM for COX-1 and 72.7 µM for COX-2). Potently blocks aspirin inactivation of COX-1 (EC50 antagonism of 200 µM aspirin ~290 nM for ovine COX-1). Decreases the secretion of total Aβ (Amyloid β40&42) by human neuronal cells and offers neuroprotection against glutamate-, nitric oxide- and superoxide-induced damage. Activates peroxisome proliferator activated receptors α and γ in both CV-1 and C3H10T1/2 cells (~100 µM - 500 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
COX-1
Product competes with ATP.
Reversible: yes
Target IC50: 4.85 µM against COX-1

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Asanuma, M., et al. 2001. J. Neurochem.76, 1895.
Blasko, I., et al. 2001. Neurobiol. Dis.8, 1094.
Ouellet, M., et al. 2001. Proc. Natl. Acad. Sci. USA98, 14583.
Casper, D., et al. 2000. Neurosci. Lett.289, 201.
Lambat, Z., et al. 2000. Metab. Brain Dis.15, 249.
Lim, G.P., et al. 2000. J. Neurosci.20, 5709.
Ogawa, O., et al. 2000. Eur. J. Pharmacol.408, 137.
Wyss-Coray, T., and Mucke, L. 2000. Nat. Med.6, 973.
Lehmann, J.M., et al. 1997. J. Biol. Chem.272, 3406.
Boneburg, E.M., et al. 1996. J. Clin. Pharmacol.36, 16S.
Mitchell, J.A., et al. 1994. Proc. Natl. Acad. Sci. USA90, 11693.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

O Ogawa et al.
European journal of pharmacology, 408(2), 137-141 (2000-11-18)
Recent studies show that a mononuclear phagocyte lineage, including microglia, plays a possible role in the pathogenesis of Alzheimer's disease through nitric oxide (NO)-mediated neurotoxicity. Epidemiological studies show that nonsteroidal anti-inflammatory drugs (NSAIDs) have a protective effect against Alzheimer's disease.
J M Lehmann et al.
The Journal of biological chemistry, 272(6), 3406-3410 (1997-02-07)
Indomethacin is a non-steroidal anti-inflammatory drug (NSAID) and cyclooxygenase inhibitor that is frequently used as a research tool to study the process of adipocyte differentiation. Treatment of various preadipocyte cell lines with micromolar concentrations of indomethacin in the presence of
I Blasko et al.
Neurobiology of disease, 8(6), 1094-1101 (2001-12-14)
Trying to decrease the production of Amyloid beta (Abeta) has been envisaged as a promising approach to prevent neurodegeneration in Alzheimer's disease (AD). A chronic inflammatory reaction with activated microglia cells and astrocytes is a constant feature of AD. The
Ibuprofen, inflammation and Alzheimer disease.
T Wyss-Coray et al.
Nature medicine, 6(9), 973-974 (2000-09-06)
J A Mitchell et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(24), 11693-11697 (1993-12-15)
Constitutive cyclooxygenase (COX-1; prostaglandin-endoperoxide synthase, EC 1.14.99.1) is present in cells under physiological conditions, whereas COX-2 is induced by some cytokines, mitogens, and endotoxin presumably in pathological conditions, such as inflammation. Therefore, we have assessed the relative inhibitory effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service