Skip to Content
Merck
All Photos(1)

Key Documents

385581-M

Sigma-Aldrich

11beta-HSD1 Inhibitor, 10j

The 11beta-HSD1 Inhibitor, 10j, also referenced under CAS 1009373-58-3, controls the biological activity of 11beta-HSD1. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

11beta-HSD1 Inhibitor, 10j

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H33BrN2O3
CAS Number:
Molecular Weight:
501.46
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell permeable, spirocyclic urea that acts as a selective inhibitor of 11β-hydroxysteroid dehydrogenase 1 (11β-HSD1) (IC50 of 1.6 nM and 5.3 nM) in enzyme and human adipocyte assays, respectively. Consisting of 2 enantiomers, 10j2 (R Isomer) is shown to be ˜ 2-3X more potent than 10j1 (S Isomer), in both aforementioned assays. 11β-HSD1 activity in the presence of 10j2 (10mg/kg in animals p.o.) results in an ˜ 80% reduction in the concentration of plasma cortisol over vehicle treated controls.

Warning

Toxicity: Standard Handling (A)

Other Notes

Tice, C.M., et al. 2009. Bioorg. Med. Chem. Lett.20, 881.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service