Skip to Content
Merck
All Photos(1)

Key Documents

1.70340

Supelco

Phosphorus ICP standard

traceable to SRM from NIST H₃PO₄ in H₂O 1000 mg/l P Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

suitable for EPA 200.7

product line

Certipur®

concentration

1000 mg/L P in H2O

technique(s)

ICP: suitable

pH

1 (20 °C in H2O)

density

1.000 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Phosphorus Standard can be used as a calibration standard in the determination ofphosphorus in protein samples by inductively coupled plasma quadrupole mass spectrometry (ICP-QMS)
The phosphorus ICP standard may be used as following:
  • Estimation of metals and phosphorus in vegetable seeds by ICP-OES and ICP-MS
  • Inductively coupled plasma quadrupole-based mass spectrometry (ICP-QMS) based quantification of phosphorus in protein samples
  • Development and validation of an inductively coupled plasma -optical emission spectroscopy (ICP-OES) method for the determination of rare earth elements, thorium, yttrium, and phosphorus in monazite samples
  • Comparative analysis of inductively coupled plasma-mass spectrometry (ICP-MS) and colorimetry for the determination of total and extractable phosphorus in soils
  • Electrochemical measurement of dissolved phosphorus in canal water samples
  • Determination of phosphorus and potassium content of commercial inorganic fertilizers by ICP-OES

Analysis Note

1000 mg/L P in water, prepared with high-purity H3PO4 and water.

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method validation and uncertainty for the determination of rare earth elements, yttrium, thorium and phosphorus in monazite samples by ICP-OES
Padmasubashini V, et al.
Chemistry International, 6(3), 98-109 (2020)
Metals and phosphorus determination in vegetable seeds used in the production of biodiesel by ICP OES and ICP-MS
Chaves ES, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 96(1), 71-76 (2010)
Rapid and portable electrochemical quantification of phosphorus
Kolliopoulos A, et al.
Analytical Chemistry, 87(8), 4269-4274 (2015)
Protein phosphorylation stoichiometry by simultaneous ICP-QMS determination of phosphorus and sulfur oxide ions: a multivariate optimization of plasma operating conditions
Ciavardelli D, et al.
Talanta, 80(4), 1513-1525 (2010)
Comparison of inductively coupled plasma mass spectrometry and colorimetric determination of total and extractable phosphorus in soils
Ivanov K, et al.
Spectrochimica Acta. Part B: Atomic Spectroscopy, 71, 117-122 (2012)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service