Skip to Content
Merck
All Photos(1)

Key Documents

124011

Sigma-Aldrich

Akt Inhibitor IV

The Akt Inhibitor IV, also referenced under CAS 681281-88-9, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Akt Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H27IN4S
CAS Number:
Molecular Weight:
614.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[I-].[s]1c2c(nc1c3cc4[n](c([n+](c4cc3)c6ccccc6)C=CN(C)c5ccccc5)CC)cccc2

InChI

1S/C31H27N4S.HI/c1-3-34-28-22-23(31-32-26-16-10-11-17-29(26)36-31)18-19-27(28)35(25-14-8-5-9-15-25)30(34)20-21-33(2)24-12-6-4-7-13-24;/h4-22H,3H2,1-2H3;1H/q+1;/p-1

InChI key

NAYRELMNTQSBIN-UHFFFAOYSA-M

General description

A cell permeable and reversible benzimidazole compound that inhibits Akt phosphorylation/activation by targeting the ATP binding site of a kinase upstream of Akt, but downstream of PI 3-K. Shown to block Akt-mediated FOXO1a nuclear export (IC50 = 625 nM) and cell proliferation (IC50<1.25 M) in 786-O cells. Unlike phosphatidylinositol analog-based Akt inhibitors (Cat. Nos. 124005, 124008, 124009), this inhibitor does not affect PI3K. A 10 mM (1 mg/163 l) solution of Akt Inhibitor IV in DMSO is also available.
A cell-permeable and reversible benzimidazole compound that inhibits Akt phosphorylation/activation presumably by targeting the ATP binding site of a kinase upstream of Akt, but downstream of PI3-K. Shown to block Akt-mediated FOXO1a nuclear export (IC50 = 625 nM) and cell proliferation (IC50<1.25 µM) in 786-O cells. Unlike phosphatidylinositol analog-based Akt inhibitors (Cat. Nos. 124005, 124008, 124009), this inhibitor does not affect PI3-K.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Akt
Product does not compete with ATP.
Reversible: yes
Target IC50: 625 nM in blocking Akt-mediated FOXO1a nuclear export; <1.25 µM in blocking Akt-mediated cell proliferation in 786-O cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kau, T.R., et al. 2003. Cancer Cell4, 463.



Selected Citations
Lee, J., et al. 2009. Cell Stem Cell5, 76.


Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mengping Zhang et al.
Journal of cellular physiology, 233(6), 4748-4758 (2017-11-18)
While overexpression of FSCN1 is reported in several cancers, the prognostic significance of FSCN1 in renal cell carcinoma (RCC) and the molecular mechanisms involved remain largely unclear. We retrospectively enrolled 194 patients with non-metastatic clear-cell RCC undergoing nephrectomy in our
Oleksii S Rukhlenko et al.
Nature, 609(7929), 975-985 (2022-09-15)
Understanding cell state transitions and purposefully controlling them is a longstanding challenge in biology. Here we present cell state transition assessment and regulation (cSTAR), an approach for mapping cell states, modelling transitions between them and predicting targeted interventions to convert
Bing Xu et al.
Frontiers in neuroscience, 14, 766-766 (2020-08-28)
Poor postoperative pain (POP) control increases perioperative morbidity, prolongs hospitalization days, and causes chronic pain. However, the specific mechanism(s) underlying POP is unclear and the identification of optimal perioperative treatment remains elusive. Akt and mammalian target of rapamycin (mTOR) are
Michael Xiao et al.
eLife, 12 (2023-04-14)
Quiescent stem cells are activated in response to a mechanical or chemical injury to their tissue niche. Activated cells rapidly generate a heterogeneous progenitor population that regenerates the damaged tissues. While the transcriptional cadence that generates heterogeneity is known, the
Roshini Fernando et al.
Journal of immunology (Baltimore, Md. : 1950), 204(7), 1724-1735 (2020-02-23)
IL-23 and IL-12, two structurally related heterodimeric cytokines sharing a common subunit, divergently promote Th cell development and expansion. Both cytokines have been implicated in the pathogenesis of thyroid-associated ophthalmopathy (TAO), an autoimmune component of Graves disease. In TAO, CD34+

Articles

AldeRed™ 588-A detects ALDH activity in cancer stem cells, aiding in cancer research.

AldeRed™ 588-A detects ALDH activity in cancer stem cells, aiding in cancer research.

AldeRed™ 588-A detects ALDH activity in cancer stem cells, aiding in cancer research.

AldeRed™ 588-A detects ALDH activity in cancer stem cells, aiding in cancer research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service