1.18771
Fluoride Test Kit, colorimetric
0-0.8 mg/L (F), MQuant®
Synonym(s):
Fluoride Visual Test Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116116
NACRES:
NA.24
Recommended Products
Product Name
Fluoride Test, colorimetric, 0-0.8 mg/L (F), for use with MCOLORTEST®
specific analyte(s)
fluoride
Quality Level
measuring range
0-0.8 mg/L (F)
compatibility
for use with MCOLORTEST®
detection method
colorimetric
storage temp.
15-25°C
General description
Method: colorimetric with color card 0 - 0.15 - 0.3 - 0.5 - 0.8 mg/l F MQuant®
This MQuant® Fluoride test (formally MColortest®) is suitable for the colorimetric determination using the easy to use color card. The reagents are added to the sample, resulting in a color reaction. The concentration is determined by allocating the color to a value on a reference scale.
Analysis Note
Application test: conforms
Legal Information
MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service