Skip to Content
Merck
All Photos(1)

Key Documents

1.14821

Supelco

Gold Test, photometric

photometric, 0.5-12.0 mg/L (Au), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Gold Test, photometric, 0.5-12.0 mg/L (Au), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 75 tests
75 tests

specific analyte(s)

gold

measuring range

0.5-12.0 mg/L (Au)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Rhodamine B)

storage temp.

15-25°C

General description

Method: photometric 0.5 - 12.0 mg/l Au Spectroquant®

This Spectroquant® Gold Reagent Test allows the accurate determination of the gold ion concentration in aqueous samples. To analyze undissolved or complex-bound gold the sample has to be decomposed by digestion.


Method applied: All gold ions are oxidized to gold(III) ions by an oxidizing agent. In sulfuric solution these ions react with rhodamine B to form a red-violet complex that is extracted and subsequently determined photometrically.


The Spectroquant®Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.


All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service