Skip to Content
Merck
All Photos(1)

Key Documents

1.14776

Supelco

Nitrite Test, photometric

photometric, 0.002-1.00 mg/L (NO2-N), 0.007-3.28 mg/L (NO2-), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Nitrite Test, photometric, 0.002-1.00 mg/L (NO2-N), 0.007-3.28 mg/L (NO2-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 1000 tests
sufficient for 335 tests (depending on package size)

specific analyte(s)

nitrite

reaction suitability

reaction type: Griess reaction

measuring range

0.002-1.00 mg/L (NO2-N)
0.007-3.28 mg/L (NO2-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Griess’ reaction)

storage temp.

15-25°C

General description

Method: photometric 0.002 - 1.00 mg/l NO₂-N 0.007 - 3.28 mg/l NO₂⁻ Spectroquant®
The method is analogous to EPA 354.1, APHA 4500-NO2- B, and DIN EN 26 777.
This Spectroquant® Nitrate Cell Test allows the accurate quantification of nitrate in water samples. Nitrate is an essential nutrient required for the growth of animals and plants. However, elevated levels of nitrate can be a health risk, in particular for pregnant women and infants. Therefore, nitrate levels in water sources are regulated in many countries.
Method applied: In acidic solution nitrite ions react with sulfanilic acid to form a diazonium salt that reacts with N-(1-naphthyl)ethylenediamine dihydrochloride to form a red-violet azo dye which is determined photometrically.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Photometric determination using the Griess method subsequent to extraction

Photometric determination of nitrite ions in soil samples by Griess method using Spectroquant® test kits and spectrophotometer.

Photometric determination of nitrite ions in compost by Griess method using Spectroquant®

Photometric determination of nitrite ions in compost by Griess method using Spectroquant®

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service