Skip to Content
Merck
All Photos(1)

Key Documents

1.14561

Supelco

Cyanide Test, photometric

photometric, 0.010-0.500 mg/L (CN-)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Pricing and availability is not currently available.

Product Name

Cyanide Cell Test, photometric, 0.010-0.500 mg/L (CN-)

usage

sufficient for 25 tests

Quality Level

specific analyte(s)

cyanide

measuring range

0.010-0.500 mg/L (CN-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Barbituric acid, pyridine-carboxylic acid)

storage temp.

15-25°C

General description

The method corresponds to DIN 38405-13 and is analogous to ISO 6703, EPA 335.2, APHA 4500-CN-E, and ASTM D2036-09 D.


This Spectroquant® Cyanide Cell Test allows the accurate quantification of the free and readily liberated cyanide content in aqueous samples.


Method applied: Cyanide ions react with a chlorinating agent to form cyanogen chloride which together with 1,3-dimethylbarbituric acid forms a violet dye (pyridine-free König reaction) that is determined photometrically.


The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.


All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service