1.09410
ICP Multi element standard solution XXIII
for mass calibration Certipur®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
Recommended Products
Quality Level
product line
Certipur®
technique(s)
ICP: suitable
pH
<1 (20 °C in H2O)
density
1.017 g/m3 at 20 °C
format
multi-component solution
storage temp.
15-25°C
Analysis Note
Ba (barium): 0.5 - 2.0 ng/ml
B (boron): 0.5 - 2.0 ng/ml
Co (cobalt): 0.5 - 2.0 ng/ml
Fe (iron): 0.5 - 2.0 ng/ml
Ga (Gallium): 0.5 - 2.0 ng/ml
In (Indium): 0.5 - 2.0 ng/ml
K (potassium): 0.5 - 2.0 ng/ml
Li (Lithium): 0.5 - 2.0 ng/ml
Lu (Lutetium): 0.5 - 2.0 ng/ml
Na (sodium): 0.5 - 2.0 ng/ml
Rh (Rhodium): 0.5 - 2.0 ng/ml
Sc (Scandium): 0.5 - 2.0 ng/ml
Tl (Thallium): 0.5 - 2.0 ng/ml
U (Uranium): 0.5 - 2.0 ng/ml
y (Yttrium): 0.5 - 2.0 ng/ml
B (boron): 0.5 - 2.0 ng/ml
Co (cobalt): 0.5 - 2.0 ng/ml
Fe (iron): 0.5 - 2.0 ng/ml
Ga (Gallium): 0.5 - 2.0 ng/ml
In (Indium): 0.5 - 2.0 ng/ml
K (potassium): 0.5 - 2.0 ng/ml
Li (Lithium): 0.5 - 2.0 ng/ml
Lu (Lutetium): 0.5 - 2.0 ng/ml
Na (sodium): 0.5 - 2.0 ng/ml
Rh (Rhodium): 0.5 - 2.0 ng/ml
Sc (Scandium): 0.5 - 2.0 ng/ml
Tl (Thallium): 0.5 - 2.0 ng/ml
U (Uranium): 0.5 - 2.0 ng/ml
y (Yttrium): 0.5 - 2.0 ng/ml
Legal Information
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service